|
The traditional process of drug discovery based on natural
secondary metabolites has often been slow, costly, and labor-intensive. Even with the advent of combinatorial chemistry and high-throughput screening in the past two decades, the generation of leads is dependent on the reliability of the individual reactions to
construct the new molecular framework.
Click chemistry is a newer approach to the synthesis of
drug-like molecules that can accelerate the drug discovery process by utilizing
a few practical and reliable reactions. Sharpless and coworkers defined what makes a click reaction as one that is wide in scope and easy to perform, uses only readily available reagents, and is insensitive to oxygen and water. In
fact, in several instances water is the ideal reaction solvent, providing the
best yields and highest rates. Reaction work-up and purification uses benign
solvents and avoids chromatography.1
Of the reactions comprising the click universe, the
“perfect” example is the Huisgen 1,3-dipolar cycloaddition
of alkynes to azides to form 1,4-disubsituted-1,2,3-triazoles (Scheme 1). The
copper(I)-catalyzed reaction is mild and very efficient, requiring no
protecting groups, and requiring no purification in many cases.2The azide and alkyne functional groups are largely inert towards biological molecules and aqueous environments, which allows the use of the Huisgen 1,3-dipolar cycloaddition in target guided
synthesis3 and activity-based protein profiling.4 The triazole has similarities to the ubiquitous amide moiety found in nature, but unlike amides, is not susceptible to cleavage. Additionally, they are
nearly impossible to oxidize or reduce.
Using Cu(II) salts with ascorbate has been the method of
choice for preparative synthesis of 1,2,3-triazoles, but is problematic in
biocojugation applications. However, tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, TBTA (Figure 1), has been shown to effectively enhance the copper-catalyzed cycloaddition without damaging biological scaffolds.5
Recently, Sharpless and coworkers reported the
ruthenium-catalyzed cycloaddition of azides to alkynes to form the
complementary 1,5-disubstituted triazoles.6 Several ruthenium complexes were employed, but the pentamethylcyclopentadienyl (Cp*) analogues gave the best results, with
Cp*RuCl(PPh3)2 being employed in most cases. Whereas the Cu(I)-catalyzed reaction is limited to terminal alkynes, the Ru(II)-catalyzed reaction is active with internal alkynes as well (Scheme 2).
Of course many aliphatic azides are not commercially available. Carreira and coworkers
recently reported the hydroazidation of unactivated olefins to yield alkyl
azides in the presence of a cobalt catalyst prepared in situ from a Schiff base
ligand and Co(BF4)2·6H2O (Scheme 3).7 Additionally, the reaction can be coupled to the Sharpless cycloaddition to yield the 1,4-triazole in a one-pot process.
Sigma-Aldrich is pleased to offer these reagents and substrates for your research requirements in the exciting field of click chemistry.
Product Information
| Product # |
Product Name |
Add to Cart |
|
Click Catalyst and Ligands |
| C1297 |
Copper(II) sulfate, ReagentPlus®, ≥99% |
|
|
209198 |
Copper(II) sulfate pentahydrate
ACS reagent, ≥98.0% |
|
|
326755 |
Copper(II) acetate, 98% |
|
|
205540 |
Copper(I) iodide, 98% |
|
|
212865 |
Copper(I) bromide, 98% |
|
|
A7631 |
(+)-Sodium L-ascorbate crystalline, ≥98% |
|
|
673293 |
Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride |
|
|
678937 |
tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, TBTA |
|
|
Hydroazidation of Olefins |
|
676551 |
Potassium 2-(3,5-di-tert-butyl-2-hydroxybenzylideneamino)-2,2-diphenylacetate,
95% |
|
|
399957 |
Cobalt(II) tetrafluoroborate hexahydrate, 99% |
|
|
Azide Sources |
|
480525 |
Lithium azide solution, 20 wt. % in H2O |
|
|
438456 |
Sodium azide, 99.99+ % |
|
|
155071 |
Azidotrimethylsilane, 95% |
|
|
178756 |
Diphenyl phosphoryl azide, 97% |
|
|
349488 |
Azidotrimethyltin(IV), 97% |
|
|
651664 |
Tetrabutylammonium azide, 97% |
|
|
PEG Azides |
|
17758 |
11-Azido-3,6,9-trioxaundecan-1-amine, technical,
≥90% (GC) |
|
|
76172 |
O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene
glycol, ≥90% (oligomer purity) |
|
|
76318 |
O-(2-Aminoethyl)-O′-(2-azidoethyl)heptaethylene
glycol, ≥90% (oligomer purity) |
|
|
77787 |
O-(2-Aminoethyl)-O′-(2-azidoethyl)nonaethylene
glycol , ≥90% (oligomer purity) |
|
|
71613 |
O-(2-Azidoethyl)-O-[2-(diglycolyl-amino)ethyl]heptaethylene glycol, ≥90% (oligomer
purity) |
|
|
78691 |
O-(2-Azidoethyl)-O′-[2-(diglycolyl-amino)ethyl]undecaethylene glycol, ≥90% (oligomer
purity) |
|
|
Organic Azides |
|
59955 |
α-Azidoisobutyric acid solution, ~15% in heptane
(T) |
|
|
52916 |
α-Azidoisobutyric acid solution, purum, ~15% in
heptane (T) |
|
|
77213 |
Fluka Ethyl azidoacetate solution, purum, ~25%
in toluene (NMR) |
|
|
93528 |
Ethyl azidoacetate solution, purum, ~25% in
ethanol (NMR) |
|
|
244546 |
Azidomethyl phenyl sulfide, 95% |
|
|
359556 |
4-Azidoaniline hydrochloride, 97% |
|
|
359564 |
4-Azidophenyl isothiocyanate, 97% |
|
|
276219 |
1-Azidoadamantane, 97% |
|
|
514004 |
1-Azido-1-deoxy-β-D-glucopyranoside |
|
|
513989 |
1-Azido-1-deoxy-β-D-galactopyranoside, 97% |
|
|
M6691 |
α-D-Mannopyranosyl azide |
|
|
152854 |
4-Methoxybenzyloxycarbonyl azide, 95% |
|
|
340138 |
4-Carboxybenzenesulfonazide, 97% |
|
|
A6057 |
4-Azidophenacyl bromide |
|
|
11550 |
4-Azidophenacyl bromide, BioChemika, ≥98.0% (HPLC) |
|
|
404764 |
4-Acetamidobenzenesulfonyl azide, 97% |
|
|
A4810 |
3′-Azido-2′,3′-dideoxyuridine, ≥98% (TLC) |
|
|
A2169 |
3′-Azido-3′-deoxythymidine, ≥98% (HPLC) |
|
|
11546 |
3′-Azido-3′-deoxythymidine, BioChemika, ≥99.0% (HPLC) |
|
|
11544 |
2′-Azido-2′-deoxyuridine, BioChemika, ≥99.0%
(N) |
|
|
573213 |
(4S)-4-[(1R)-2-Azido-1-(benzyloxy)ethyl]-2,2-dimethyl-1,3-dioxolane |
|
|
31755 |
7-(Diethylamino)coumarin-3-carbonyl azide,
BioChemika, for fluorescence, ≥95% (HPLC) |
|
|
493406 |
[3aS-(3aα,4α,5β,7aα)]-5-Azido-7-bromo-3a,4,5,7a-tetrahydro-2,2-dimethyl-1,3-benzodioxol-4-ol,
99% |
|
|
33486 |
Bis(3-azidophenyl) sulfone, purum, ≥97.0% (HPLC) |
|
|
497029 |
3-Azido-2,3-dideoxy-1-O-(tert-butyldimethylsilyl)-β-D-arabinohexopyranose |
|
|
11549 |
5-Azido-2-nitrobenzoic acid N-hydroxysuccinimide
ester |
|
|
A0456 |
(2S,3R,4E)-2-Azido-4-octadecene-1,3-diol |
|
|
R109 |
Ro 15-4513 |
|
|
514012 |
1-Azido-1-deoxy-β-D-lactopyranoside |
|
|
A1262 |
8-Azidoadenosine 3′:5′-cyclic monophosphate,
~95% |
|
|
283029 |
2,6-Bis(4-azidobenzylidene)-4-methylcyclohexanone, 97% |
|
|
513970 |
1-Azido-1-deoxy-β-D-galactopyranoside
tetraacetate, 97% |
|
|
513997 |
1-Azido-1-deoxy-β-D-glucopyranoside tetraacetate |
|
|
G4168 |
α-D-Mannopyranosyl azide tetraacetate, ≥90%
(TLC) |
|
|
A3407 |
6-(4-Azido-2-nitrophenylamino)hexanoic acid N-hydroxysuccinimide
ester |
|
|
E2028 |
Ethidium bromide monoazide, ≥95% (HPLC) |
|
|
510947 |
1-O-tert-Butyldimethylsilyl
2-azido-2-deoxy-β-D-glucopyranoside 3,4,6-triacetate |
|
|
363227 |
4,4'-Diazido-2,2'-stilbenedisulfonic acid
disodium salt hydrate |
|
|
A6830 |
8-Azido-cyclic adenosine diphosphate-ribose,
≥95% (HPLC), lyophilized powder |
|
|
79728 |
Photobiotin acetate, BioChemika, ≥95% (HPLC) |
|
|
56385 |
Photobiotin acetate, BioChemika, puriss., ≥98.0%
(TLC) |
|
References:
- For recent reviews, see: (a) Kolb, H. C.; Sharpless, K. B. Drug Discovery Today 2003, 8, 1128. (b)Kolb, H. C. et al. Angew. Chem. Int. Ed. 2001, 40, 2004.
- (a) Rostovtsev, V. V. et al. Angew. Chem.
Int. Ed.2002, 41, 2596. (b) Tornøe, C. W. et al. J. Org.
Chem. 2002, 67, 3057.
- (a) Manetsch, R. et al. J. Am. Chem. Soc.2004,
126, 12809. (b) Lewis, W. G. et al.Angew. Chem. Int. Ed.
2002, 41, 1053.
- Speers, A. E. J. Am. Chem. Soc. 2003,
125, 4686.
- Chan, T.R. et al. Org. Lett 2004,6, 2853.
- Zhang, L. et al. J. Am. Chem. Soc. 2005,127, 15998.
- Waser, J. et al. J. Am. Chem. Soc. 2005,127, 8294.
|