Evans-Type Oxazolidinethione And Thiazolidinethione Auxiliaries
The
asymmetric aldol reaction mediated by chiral auxiliaries is considered to be one
of the most important methods for asymmetric C-C bond formation. Chiral
oxazolidinethiones and thiazolidinethiones, structural variants of the
well-known Evans oxazolidinones, have proven to be highly selective and
efficient chiral auxiliaries.1 The acetate aldol reaction of
titanium enolates of the N-acylated auxiliaries with aldehydes results in
excellent diastereoselectivities. Interestingly, the syn aldol product
was obtained in high diastereomeric ratio when using 1 equivalent of sparteine,
while the anti aldol product was obtained preferentially when 2
equivalents of base were employed (Scheme 1).2 However, the
corresponding oxazolidinones are only able to achieve poor
diastereoselectivities in this particular transformation.3
In a series of experiments,
Crimmins has shown in detail that by choosing the appropriate reaction
conditions it is possible to selectively synthesize aldol condensation products
bearing either “Evans-syn” or “non-Evans-syn” stereochemistry
starting with the same chiral auxiliary (Scheme 2)! Both N-propionyl
oxazolidinethiones and N-propionyl thiazolidinethiones can be used to
this effect. The change in facial selectivity in the aldol additions is
proposed to be a result of switching between chelated and nonchelated transition
states in the mechanistic pathway.4
Iterative aldol sequences with high diastereoselectivity can also be
accomplished. Crimmins demonstrated the utility of this methodology in an
iterative aldol sequence giving access to either the syn-syn-syn adduct
or syn-anti-syn adduct depending on the reaction conditions (Scheme 3).
In
contrast to the oxazolidinone analogs, the N-acyl thiazolidinethiones and
N-acyl oxazolidinethiones can be directly reduced to their corresponding
aldehydes and the chiral auxiliary by reductive cleavage with diisobutylaluminum
hydride.5 Alcoholysis using methanol and imidazole leads to the
corresponding esters, while transamination to the Weinreb amides can be
accomplished by treatment with N,O-dimethylhydroxylamine
hydrochloride in the presence of imidazole4 or organoaluminum
compounds (Scheme 4).6
Recently, the thiazolidinethione auxiliary was successfully applied by Crimmins
and DeBaillie in a convergent enantioselective total synthesis of Bistramide A,
a member of a new class of bioactive molecules isolated from the marine ascidian
Lissoclinum bistratum with neuro- and cytotoxic properties as well as
effects on cell cycle regulation. The pyran ring fragment was constructed
diastereoselectively in a total of 10 steps starting from the TIPS-protected
aldehyde and the chlorotitanium enolate of (4S)-N-propionyl-4-benzylthiazolidine-2-thione
(Scheme 5). This stereoinducing step proceeded in excellent yield (87%) and gave
a diastereomeric ratio of >98:2.7
|
Product # |
Product Name/Description |
Structure |
Add to Cart |
|
39933 |
(S)-4-Isopropyl-1,3-thiazolidine-2-thione |
 |
|
|
05329 |
(R)-4-Isopropyl-1,3-thiazolidine-2-thione
|
39911 |
(S)-4-Phenyl-1,3-thiazolidine-2-thione
|
05802 |
(R)-4-Phenyl-1,3-thiazolidine-2-thione
|
06357 |
(S)-4-Benzyl-1,3-thiazolidine-2-thione
|
42787 |
(R)-4-Benzyl-1,3-thiazolidine-2-thione
|
04987 |
(S)-4-Isopropyl-1,3-oxazolidine-2-thione
|
08914 |
(R)-4-Isopropyl-1,3-oxazolidine-2-thione
|
08913 |
(S)-4-Phenyl-1,3-oxazolidin-2-thione
|
00762 |
(R)-4-Phenyl-1,3-oxazolidine-2-thione
|
08416 |
(S)-4-Benzyl-1,3-oxazolidine-2-thione
|
00749 |
(R)-4-Benzyl-1,3-oxazolidine-2-thione
|
 |
|
References:
(1) Velàzquez, F.; Olivo, H. F. Curr. Org. Chem. 2002, 6, 303.
(2) Hodge, M. B.; Olivo, H. F. Tetrahedron 2004, 69, 9397.
(3) Evans, D. A.; Bartroli, J.; Shih, T. L. J. Am. Chem. Soc. 1981, 103, 2127.
(4) Crimmins, M. T. et al. J. Org. Chem. 2001, 66, 894.
(5) Crimmins, M. T.; Chaudhary, K. Org. Lett. 2000, 2, 775.
(6) Paquette, L. A.; Braun, A. Tetrahedron Lett. 1997, 38, 5119.
(7) Crimmins,
M. T.; DeBaillie, A. C. J. Am. Chem. Soc. 2006, 128, 4936.
|