Metal-free Asymmetric Catalysis
Developed by Professor David MacMillan at Caltech, imidazolidinone-based OrganoCatalysts™ are designed to serve as general catalysts for a variety of asymmetric transformations. The first highly enantioselective organocatalytic Diels-Alder reaction using (5S)-2,2,3-trimethyl-5-phenylmethyl-4-imidazolidinone monohydrochloride was reported by MacMillan in his pioneering work in 2000 (Scheme 1).1 The activated iminium ion, formed through condensation of the imidazolidinone and an α,β-unsaturated aldehyde, underwent reaction with various dienes to yield [4+2]-cycloadducts in excellent yields and enantioselectivities.
Other organocatalytic transformations such as 1,3-dipolar cycloadditions,2 Friedel-Crafts alkylations,3 α-chlorinations,4 α-fluorinations,5 and intramolecular Michael reactions6 were reported using MacMillan’s Imidazolidinone OrganoCatalysts™, all proceeding with high levels of enantioselectivity (Scheme 2).
MacMillan found an optimized structure in (2S,5S)-(−)-2-tert-butyl-3-methyl-5-benzyl-4-imidazolidinone for the Friedel-Crafts alkylation of indoles (Scheme 3). 6,7
The synthetic utility of this concept was later demonstrated in the total synthesis of
(–)-flustramine B, a biologically active pyrroloindoline-containing alkaloid (Scheme 4).
Sigma-Aldrich, in collaboration with Materia, Inc., is pleased to offer the following six imidazolidinone OrganoCatalysts™.
|
Product # |
Product Name/Description |
Structure |
Add to Cart |
|
674575 |
MacMillan Imidazolidine OrganoCatalyst™ Kit I |
- |
|
|
661910 |
(R)-(-)-2-(tert-Butyl)-3-methyl-4-oxoimidazolidinium trifluoroacetate |
 |
|
|
661902 |
(S)-(+)-2-(tert-Butyl)-3-methyl-4-oxoimidazolidinium trifluoroacetate |
 |
|
|
668842 |
(2R,5R)-(+)-5-Benzyl-3-methyl-2-(5-methyl-2-furyl)-2-imidazolidinone |
 |
|
|
668540 |
(2S,5S)-(-)-5-Benzyl-3-methyl-2-(5-methyl-2-furyl)-2-imidazolidinone |
 |
|
|
569763 |
(5S)-2,2,3-Trimethyl-5-phenylmethyl-4-imidazolidinone monohydrochloride |
 |
|
|
663069 |
(5R)-2,2,3-Trimethyl-5-phenylmethyl-4-imidazolidinone monohydrochloride |
 |
|
|
663107 |
(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone |
 |
|
|
663093 |
(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone |
 |
|
|
663077 |
(5R)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone dichloroacetic acid |
 |
|
|
663085 |
(5S)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone dichloroacetic acid |
 |
|
References:
1. Ahrendt, K. A.; Borths, C. J.; MacMillan, D. W. C. J. Am. Chem. Soc. 2000, 122, 4243.
2. Jen, W. S.; Wiener, J. J. M.; MacMillan, D. W. C. J. Am. Chem. Soc. 2000, 122, 9874.
3. Paras, N. A.; MacMillan, D. W. C. J. Am. Chem. Soc. 2001, 123, 4379-4371.
4. Brochu, M. P.; Brown, S. P.; MacMillan, D. W. C. J. Am. Chem. Soc. 2004, 126, 4108.
5. Beeson, T. D.; MacMillan, D. W. C. J. Am. Chem. Soc. 2005, 127, 8826.
6. Fonseca, M. H.; List, B. Angew. Chem. Int. Ed. 2004, 43, 3958.
7. Austin, J. F.; MacMillan, D. W. C. J. Am. Chem. Soc. 2002, 124, 1172.
OrganoCatalysts™ is a trademark of Materia, Inc.
|