Chemical Synthesis
Fluorous
Product Catalog
Technology Spotlights
ChiroSolv Kits
APhos
Chiral Phosphoric Acids
Takasago Ligands
catASium
cataCXium
Key Resources
Drug Discovery
Materials Science
Stable Isotopes (ISOTEC)
 Umicore Precatalysts for Asymmetric Reactions
Chemical Synthesis
Precatalysts for Asymmetric and Cross-Coupling Catalysis
Precatalysts for Asymmetric and Cross-Coupling Catalysis
Introduction
Sigma-Aldrich and Umicore are happy to offer a series of precatalysts for asymmetric catalysis and Pd based cross-coupling catalysis. Our partnership with Umicore allows us to offer a portfolio of metal complexes with batch-to-batch consistency for a plethora of catalytic reactions. Twenty different precatalyst complexes are available, providing access to a hundred different catalytic reactions.
Representative Application
Catalytic hydrogenation is a powerful and effective method used for the reduction of organic molecules. Krische and co-workers have developed methods to utilize this catalytic reaction to generate C–C coupling. After extensive studies done using Rhodium as a precatalyst, Krische and co-workers realized the limitations of Rhodium to catalyze C–C bond forming hydrogenation of only conjugated alkynes. To circumvent these limitations, the utilization of another catalyst was necessary. To expand the scope, Ngai et al. investigated the potency of Ir(COD)2BARF for this transformation. These new complexes proved to be very active with nonconjugated, alkyl-substituted alkynes. In a typical reaction,
Ir(COD)2BARF, DPPF ligand and triphenylacetic acid are used to catalyze the coupling hydrogenation of 3-hexyne with a variety of α-ketoesters to yield α-hydroxyesters in good yields.
Precatalysts Scheme 1
Reference: Ngai, W.-Y. et al. J. Am. Chem. Soc. 2007, 129, 280.
Scheme 1: Iridium catalyzed C–C bond forming hydrogenation
Product Information
Product # Product Name Structure Add to Cart
683353 Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) Shopping Cart
693774Bis(cyclooctadiene)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)Bis(cyclooctadiene)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)Shopping Cart
683086(Acetylacetonato)(1,5-cyclooctadiene)iridium(I)(Acetylacetonato)(1,5-cyclooctadiene)iridium(I)Shopping Cart
683094Bis(1,5-cyclooctadiene)diiridium(I) dichlorideBis(1,5-cyclooctadiene)diiridium(I) dichlorideShopping Cart
683108(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)Shopping Cart
683116(Acetylacetonato)(norbornadiene)rhodium(I)(Acetylacetonato)(norbornadiene)rhodium(I)Shopping Cart
683132Chloro(1,5-cyclooctadiene)rhodium(I) dimerChloro(1,5-cyclooctadiene)rhodium(I) dimerShopping Cart
683140Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborateBis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborateShopping Cart
683159Bis(1,5-cyclooctadiene)rhodium(I) trifluorosulfonateBis(1,5-cyclooctadiene)rhodium(I) trifluorosulfonateShopping Cart
683167Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimerBicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimerShopping Cart
683175Bis(norbornadiene)rhodium(I) tetrafluoroborateBis(norbornadiene)rhodium(I) tetrafluoroborateShopping Cart
686891Pentamethylcyclopentadienylrhodium(III) chloride dimerPentamethylcyclopentadienylrhodium(III) chloride dimerShopping Cart
683183(Acetylacetonato)dicarbonylrhodium(I)(Acetylacetonato)dicarbonylrhodium(I)Shopping Cart
683191Dichloro(1,5-cyclooctadiene)ruthenium(II) polymerDichloro(1,5-cyclooctadiene)ruthenium(II) polymerShopping Cart
683205Diiodo(p-cymene)ruthenium(II) dimerDiiodo(<I>p</I>-cymene)ruthenium(II) dimerShopping Cart
683213Dichloro(p-cymene)ruthenium(II) dimerDichloro(p-cymene)ruthenium(II) dimerShopping Cart
683221Benzeneruthenium(II) chloride, dimerBenzeneruthenium(II) chloride, dimerShopping Cart
683396Allylpalladium(II) chloride dimerAllylpalladium(II) chloride dimerShopping Cart
683388Palladium(II) acetylacetonatePalladium(II) acetylacetonateShopping Cart
683124Palladium(II) acetate Pd(OAc)2 Shopping Cart
683345Tris(dibenzylideneacetone)dipalladium(0) Pd2dba3 Shopping Cart
Back to Top