Introduction
Various optically active phosphine ligands incorporating a
chiral center at phosphorus display exceptional enantiosectivities in
metal-catalyzed asymmetric synthesis.1 For instance, known classes
P-chiral phosphine ligands offer good to excellent enantiocontrol in Ru- and Rh-catalyzed
hydrogenation reactions.2 The one limitation associated with these
ligands is their sensitivity to air, which has impeded widespread applicability
in bench-top chemistry. Imamoto and co-workers have addressed this deficiency
through the invention of QuinoxP*, which contains an electron-withdrawing
quinoxaline architecture.3 Aldrich, in our collaboration with Nippon
Chemical, is pleased to offer R,R-QuinoxP* for the research
market.†
Advantages of the QuinoxP* Ligands:
· QuinoxP* is not
oxidized nor epimerized at ambient conditions in air
·
Enantioselectivities are outstanding for various reaction paradigms
· Hydrogenations
proceed under mild reaction conditions
· Low catalyst
loadings yield high TONs
Representative Applications:
The reactivity profile of
these innovative, chiral ligands is covered below and highlights the impressive
breadth of valuable transformations mediated by QuinoxP*. These powerful
efficient ligands exhibit high levels of enantiocontrol in synthetic
transformations ranging from metal-catalyzed asymmetric 1,4-additions of
arylboronic acids, to enantioselective alkylative ring opening, to asymmetric
hydrogenations.3 It is worth noting that QuinoxP* is not oxidized at
the stereogenic phosphorus center on standing at ambient temperature in air for
more than 9 months.
Highly Asymmetric
Rhodium-Catalyzed Hydrogenation
Imamoto has gone to great lengths to develop
enantiomerically pure P-chiral ligands for industrially useful transformations
such as asymmetric hydrogenation. Impressively, a diverse array of prochiral
amino acid and amine substrates were hydrogenated with great efficiency to yield
highly enantiopure amine derivatives (Scheme 1). The authors carried out
these experiments at room temperature in methanol under low hydrogen pressures (3 atm). Note that all hydrogenation reactions were complete in
6 hours and with enantiomeric excesses ranging from 96 to 99.9%. Dramatic
stereochemical reversal, consistent with the results observed with the related (S,S)-tert-Bu-BisP*
ligands,4,5 was obtained when 1-acetylamino-1-adamantylethene was
hydrogenated to afford the S configuration amine with > 96%
enantioselectivity (Table 1).
Asymmetric 1,4-additions of Arylboronic Acids
Imamoto and co-workers exploited the high activity of the
QuinoxP* ligand in rhodium-catalyzed enantioselective 1,4-additions of
arylboronic acids to α,ß-unsaturated carbonyl substrates.3 High yields of the
addition products were obtained via running the reactions between 40 and 50 ºC (Scheme
2). The exceptional enantiocontrol exerted by this Rh(I)-catalyzed system
is evident when compared to the use of BINAP as the
chiral ligand.6
Asymmetric Pd-catalyzed Ring Opening
Imamoto and co-workers have also succeeded in developing a
Pd-catalyzed C-C bond-forming reaction,
which displays high enantioselectivities with both dimethyl- and diethylzinc (Scheme
3, Table 2). This alkylative ring-opening methodology entails
simply premixing PdCl2(cod) and QuinoxP* for 2 hours at room
temperature - leading to a highly
active catalyst. This catalyst system affords excellent yields of the
ring-opened products and selectivities that rival the highest reported for this
transformation. These results, when combined with the outstanding methodologies
presented above, indicate that QuinoxP* is useful for a broad variety of
asymmetric metal-catalyzed transformations.
Product Information
|
Product # |
Product Name/Description |
Structure |
Add to Cart |
|
676403 |
(R,R)-2,3-Bis(tert-butylmethylphosphino)quinoxaline |
 |
|
References:
(1) (a) Weinkauff, D. J. et al. J. Am. Chem. Soc. 1977, 99,
5946; (b) Spagnol, M. et al. Chem. Eur. J. 1997, 3, 1365;
(c) Hamada, Y. et al. Tetrahedron Lett. 1997, 38, 8961; (d)
Kurth, V. et al. Eur. J. Inorg. Chem. 1998, 597; (e) Mezzetti, A.
et al. Organometallics 1998, 17, 668; (f) Imamoto, T. et
al. J. Am. Chem. Soc. 1998, 120, 1635; (g) Mezzetti, A. et
al. Organometallics 1999, 18, 1041; (h) Imamoto, T. J.
Org. Chem. 1999, 64, 2988; (i) Imamoto, T. et al.
Tetrahedron: Asymmetry 1999, 10, 877; (j) van Leeuwen, P. W.
N. M. et al. J. Org. Chem. 1999, 64, 3996.
(2) (a) Zhang, Z. et al. J. Org. Chem.
2000, 65, 6223; (b) Tang, W. et al. J. Am.
Chem. Soc. 2003, 125, 9570; (c) Lei,
A. et al. J. Am. Chem. Soc. 2004, 126, 1626;
(d) Tang, W.; Zhang, X. Angew. Chem. Int. Ed. Engl. 2002, 41,
1612; (e) Tang, W.; Zhang, X. Org. Lett.
2002, 4, 4159; (f) Tang, W. et al. Org.
Lett. 2003, 5, 205; (g) Tang,
W. et al. Angew. Chem. Int. Ed. Engl.
2003, 42, 3509; (h) Xiao, D. et al. Org. Lett.
1999, 1, 1679; (i) Liu, D.; Zhang, X. Eur. J. Org. Chem.
2005, 646.
(3) Imamoto, T.; Sugita, K.; Yoshida, K. J. Am. Chem. Soc. 2005,127, 11934.
(4) Imamoto, T. et al. J. Am. Chem. Soc. 2000, 122,
10486.
(5) Imamoto, T. et al. J. Am. Chem. Soc. 2001, 123,
5268.
(6) (a) Miyaura, N. et al. J. Am. Chem. Soc. 1998, 120, 5579; (b) Hayashi, T. et al. Tetrahedron Lett. 1999, 40,
6957.
†QuinoxP* ligands are sold in collaboration with Nippon Chemical for the research market (US and international patents pending)
|