|
Partnering with Reaxa, Sigma-Aldrich offers a variety of transition metal-based
encapsulated catalysts for applications in areas such as Suzuki, Stille, and
Heck couplings, carbonylations, hydrogenation, hydrogenolysis, and
dihydroxylation.
Coupling Chemistry
Pd(II) EnCat™
Pd(II) EnCat™ covers a versatile range of immobilized palladium acetate
catalysts with or without activating ligands contained in a robust, porous
polyurea matrix which allow easier, faster and cleaner reaction processes to be
developed. The recyclable catalysts have demonstrated superb activity in
cross-couplings, Heck couplings, and carbonylations.
| Cleaner products |
Typically less than 10 ppm Pd in crude product |
| Cleaner waste streams |
Reduced metal losses in Pd EnCat™ processes |
| Fast, efficient processes |
EnCat™ beads filter easily |
| No plant contamination |
Metal and ligands are trapped within the beads |
| Improved processes |
High activity in many types of coupling reactions |
| Process intensification |
Can be used in batch and flow processes |
Technical Information for Download (PDF format):
1) Pd(0) and Pd(II) EnCat™ Presentation
(2.8 MB PDF)
2) Pd(II) EnCat™ Technical Flier
(485KB PDF)
3) Pd(II) EnCat™ User Guide
(72 KB PDF)
4) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers
(624 KB PDF)
Cheminar™ 1.1: Pd EnCat™ Technology (click here to view)
| Product # |
Product Name |
Co-encapsulated Ligand |
Description |
Add
to Cart |
|
644714 |
Pd(II) EnCat™ 30 |
none |
Extent of labeling:
0.4 mmol/g Pd loading |
|
|
644722 |
Pd(II) EnCat™ 40 |
none |
Extent of labeling:
0.4 mmol/g Pd loading |
|
|
644706 |
Pd(II) EnCat™ TPP30 |
PPh3 |
Extent of labeling:
0.4 mmol/g Pd loading, Pd
1.0, TPP 0.8 |
|
|
644692 |
Pd(II) EnCat™ TOTP30 |
P(o-tolyl)3 |
Extent of labeling:
0.4 mmol/g Pd
loading, Pd 1.0, TOTP 0.5 |
|
|
658693 |
Pd(II) EnCat™ BINAP30 |
rac-BINAP |
Extent of labeling: 0.39 mmol/g Pd
loading, Pd 1.0, BINAP 0.25 |
|
|
666335 |
Pd(II) EnCat™ Kit I |
— |
Contains 1 g units
of: 644692,
644706,
644714,
644722, and
658693 |
|
References:
1) Ladlow, M et al. Org. Biomol. Chem. 2003, 1, 2419.
2) Ley, S. V. et al. Org. Biomol. Chem. 2004, 2, 611.
3) Ley, S. V. et al. Chem. Commun. 2005, 2175.
4) Pilotti, P. et al. Pharm. Disc. 2005, 5(8), 32.
5) Nájera, C. et al. Tetrahedron 2005, 61, 12168.
6) Ley, S. V. Chem. Eur. J. 2006, 12, 4407.
7) Yamamoto, Y. et al. Synlett 2006, 1027.
8) Ley, S. V. et al. Chem. Commun. 2002, 1132.
9) Ley, S. V. et al. Chem. Commun. 2002, 1134.
10) Ley, S. V. et al. Synlett 2002, 1843.
11) Pears, D. A. et al. Aldrichimica Acta 2005, 38, 23.
12) Leeke, G. A. et al. Org. Process Res. Dev. 2007, 11, 144.
13) Ley, S. V. et al. Synlett 2007, 1251.
LaPCatTM
The LaPCatTM kit is an exclusive collection of six palladium and/or copper
containing perovskite heterogeneous catalysts for use in a wide range of organic
reactions. LaPCatTM catalysts demonstrate superb catalytic activity in many
coupling reactions with a wide variety of substrates.
| Catalyst |
Formulation |
Surface Area (m2/g) |
Particle Size (μm) |
Reactions |
| FP2 |
LaFe0.95Pd0.05O3 |
5.5 |
41.1 |
Suzuki |
| FP8 |
LaFe0.80Pd0.20O3 |
5.7 |
3.3 |
Suzuki |
| CoP |
LaFe0.57Co0.38Pd0.05O3 |
3.4 |
4.0 |
Suzuki, Heck, Sonogashira |
| CuP |
LaFe0.57Cu0.38Pd0.05O3 |
12 |
3.7 |
Suzuki, Sonogashira, Ullmann |
| CoCu |
La0.9Ce0.1Co0.60Cu0.40O3 |
7.9 |
5.4 |
Ullmann |
| YBCu |
YBa2Cu3O7 |
0.3 |
10.6 |
Ullmann |
As shown in the table above, the various LaPCatTM catalysts have broad
application in several important classes of carbon-carbon bond forming
reactions.
| Cleaner products |
Typically ≤10 ppm of metal contamination in crude product |
| Cleaner waste streams |
Reduced metal losses in LaPCat™ processes |
| Fast, efficient processes |
Equivalent or better than homogeneous catalysts |
| Improved processes |
Very low catalyst loadings achieved |
| Improved cost effectiveness |
LaPCat™ catalysts may be reused |
Technical Information for Download (PDF format):
1) LaPCatTM Presentation
(1.3 MB PDF)
2) LaPCatTM Technical Flier
(365 KB PDF)
3) LaPCatTM User Guide
(287 KB PDF)
4) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers
(624 KB PDF)
| Product # |
Product Name |
Description |
Add to Cart |
|
674052 |
LaPCat™ Perovskite Catalyst Kit I |
Six 100 mg samples of LaPCat™ catalysts:
FP2, FP8, CoP, CuP, CoCu, and YBCu |
|
Hydrogenation
Pd(0) EnCat™ 30NP
Pd(0) EnCat™ 30NP is a versatile polymer encapsulated nanoparticulate Pd(0)
hydrogenation and transfer hydrogenation catalyst with high chemoselectivity and
improved safety profile allowing easier, faster and cleaner processes to be
developed. Reaxa’s controlled manufacturing process produces regular
palladium(0) nanoparticles stabilized by the polymer matrix within the EnCat™
beads, ensuring that the catalyst performance is extremely reproducible from
batch to batch (c.f. palladium on carbon).
| Cleaner products |
Typically <1 ppm Pd in crude product |
| Cleaner waste streams |
Reduced metal losses in EnCat™ processes |
| Fast, efficient processes |
EnCat™ beads filter easily and can be recycled |
| No plant contamination |
Pd metal is trapped within the beads |
| High selectivity |
In hydrogenation and transfer hydrogenation reactions |
| Improved safety profile |
Less pyrophoric compared with Pd/C catalysts |
| Process intensification |
Can be used in batch and flow processes |
Technical Information for Download (PDF format):
1) Pd(0) and Pd(II) EnCat™ Presentation
(2.8 MB PDF)
2) Pd(0) EnCat™ 30NP Technical Flier
(518 KB PDF)
3) Pd(0) EnCat™ 30NP User Guide
(340 KB PDF)
4) Pd(0) EnCat™ 30NP Debenzylation Guide
(159 KB PDF)
5) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers
(624 KB PDF)
Cheminar™ 1.1: Pd EnCat™ Technology (click here to view)
| Product # |
Product Name |
Description |
Add to Cart |
|
653667 |
Pd(0) EnCat™ 30NP |
nanoparticles, extent of labeling: 0.4 mmol/g Pd loading |
|
References:
1) Ley, S. V. et al. Chem. Comm. 2003, 678.
2) Ley, S. V. et al. Beil. J. Org. Chem. 2006, 2, 15.
3) Ley, S. V. et al. Org. Lett. 2003, 5, 4665.
4) Ley, S. V. et al. Org. Biomol. Chem. 2004, 2, 160.
5) Ley, S. V. et al. Chem. Commun. 2002, 1132.
6) Pears, D. A. et al. Aldrichimica Acta 2005, 38, 23.
7) Quai, M. et al. Tetrahedron Lett. 2007, 48, 1241.
Pt(0) EnCat™ 40
Pt(0) EnCat™ 40 is a versatile hydrogenation catalyst demonstrating high
chemoselectivity and improved safety profile over platinum on carbon allowing
easier, faster and cleaner processes to be developed. Isolating the active
platinum within the polymer matrix has produced an easily-handled and safer
alternative to Pt/C. The catalyst is particularly effective for hydrogenation of
nitro groups and reductive aminations in the presence of halogens.
| Cleaner products
|
Very low levels of Pt in crude product |
| Cleaner waste streams
|
Reduced metal losses in EnCat™ process (typically <0.005% Pt loss) |
| Fast, efficient processes
|
EnCat™ beads filter easily |
| No plant contamination
|
Pt metal remains entrapped within the beads |
| High selectivity
|
In hydrogenation reactions |
| Process intensification
|
Can be used in batch and flow processes |
| Improved safety profile
|
Initial tests show no tendency of activated catalyst to ignite on exposure to air |
Technical Information for Download (PDF format):
1) Pt(0) EnCat™ 40 Presentation
(323 KB PDF)
2) Pt(0) EnCat™ 40 Technical Flier
(43 KB PDF)
3) Pt(0) EnCat™ 40 User
Guide (43 KB PDF)
4) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers
(624 KB PDF)
| Product # |
Product Name |
Description |
Add to Cart |
|
681504 |
Pt(0) EnCat™ 40 |
Wet, encapsulated nanoparticles, extent of labeling: 0.12
mmol/g Pt loading (dry basis) |
|
Dihydroxylation
Os EnCat™ 40
Osmium tetroxide (OsO4)is known to be one of the most reliable and effective
catalysts for oxidative cleavage and dihydroxylation of olefins. However, it is
very toxic, volatile and expensive, thus limits its usefulness at large-scale.
Os EnCat™ 40 effectively immobilizes and stabilizes osmium tetroxide to produce
a safer and easier-to-use catalyst for application in oxidation chemistry.
In the above picture, a vial containing Os EnCat™ 40 placed in the presence of
corn oil is unchanged after several months, whereas an equivalent sample of
volatile osmium tetroxide quickly reacts to produce black decomposition
products, indicating the stability of the encapsulated OsO4.
| Cleaner products |
Typically less than 100 ppm Os in crude product |
| Easier storage, transport and use |
Low residual OsO4 vapor over Os EnCat™ product |
| Faster, efficient processes |
Resin beads filter easily |
| High selectivity |
Can be used for asymmetric dihydroxylation |
| Reduced plant contamination |
OsO4 is trapped in the beads |
| Improved cost effectiveness |
Os EnCat™ 40 may be reused in repeat reaction cycles |
| Process intensification |
Can be used in batch and flow processes |
Technical Information for Download (PDF format):
1) Os EnCat™ 40 Technical Flier
(73 KB PDF)
2) Os EnCat™ 40 User Guide
(111 KB PDF)
3) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers
(624 KB PDF)
| Product # |
Product Name |
Description |
Add to Cart |
|
658685 |
Os EnCat™ 40 |
Extent of labeling:
0.3 mmol/g Os loading |
|
References:
1) Ley, S. V. et al. Org. Lett. 2003, 5, 185.
2) Borhan, B. et al. Tetrahedron Lett. 2006, 47, 3797.
3) Ley, S. V. et al. Org. Lett. 2003, 5, 185.
4) Ley, S. V. et al. Org. Biomol. Chem. 2003, 1, 3957.
|