|
In catalytic asymmetric
synthesis enzyme-mediated kinetic resolution is an important area. Lipases are
particularly successful in kinetic resolutions and hence are manufactured on a
commercial scale. However, conventional kinetic resolution has a maximum
theoretical yield of 50%. Dynamic kinetic resolution (DKR) circumvents this
limitation by epimerizing the slow-reacting enantiomer concurrently with the
kinetic resolution. In this way, one enantiomer of the epimerized substrate is
converted faster to the product (e.g. by transesterification) by the enzyme, and
the yield can theoretically be increased to 100% using DKR (Scheme 1).1
Diruthenium complex 1,
first reported by Shvo,2 has found numerous applications as a
versatile catalyst in organic synthesis, including the reduction of aldehydes
and ketones to alcohols, bimolecular disproportionation reaction of aldehydes to
esters, isomerization of allylic alcohols and oxidation of alcohols.3
Under thermal conditions, the Shvo catalyst
dissociates into the catalytically active 16-electron species 2 and
18-electron complex 3 (Scheme 2).
Bäckvall has successfully
used Shvo’s complex as an efficient epimerization catalyst in several enzyme
mediated DKRs.4 In the first step of the proposed epimerization
mechanism of secondary alcohols, one of the oxygens on 2 abstracts a
proton (removing the need of an external base as cocatalyst) and the ruthenium
metal acts as a hydride acceptor, subsequently leading to ketone formation. The
ketone generated is then reduced in reverse fashion, resulting in an overall
epimerization of the corresponding alcohol (Scheme 3).
The nature of acyl donor is
critical to take into account for successful chemoenzymatic DKR, and Bäckvall
and co-workers have discovered certain aryl esters are efficient acyl donors in
the DKR of several alcohols. An efficient protocol using immobilized Candida antarcticalipase B (CALB) is shown in Scheme 4. Based on the use of CALB in
combination with Shvo’s ruthenium catalyst andp-chlorophenyl
acetate as acyl donor, a number of secondary alcohols were successfully
transesterified and obtained in good to high yields and excellent
enantioselectivities (Scheme 5). In the case of the DKR of the racemic
a-hydroxy
esters, immobilized Pseudomonas cepacia lipase (PS-C) in cyclohexane was used.
The chemoenzymatic DKR was
also applied to symmetrical diols, hydroxy esters, azido alcohols, hydroxy
nitriles, halo alcohols and hydroxy phosphonates (Table 1). Similarly, using a
slightly higher amount of Shvo’s catalyst, various
b-hydroxy
esters were obtained in a tandem aldol-deracemization-transesterification
reaction sequence in good yields and enantioselectivities (Scheme 6).
|
Product # |
Product Name/Description |
Structure |
Add to Cart |
|
668281 |
1-Hydroxytetraphenylcyclopentadienyl-(tetraphenyl-2,4-cyclopentadien-1-one)-μ-hydrotetracarbonyldiruthenium(II) |
 |
|
|
12117 |
Lipase A, Candida
antarctica, CLEA |
- |
|
|
62288 |
Lipase B, Candida antarctica, recombinant from Aspergillus oryzae
|
- |
|
|
16698 |
Lipase, Candida antarctica, CLEA
|
- |
|
|
62299 |
Lipase from Candida antarctica
|
- |
|
|
73940 |
Lipase immobilized from Candida antarctica
|
- |
|
|
89137 |
Lipase, immobilized in Sol-Gel-AK on pumice from Candida antarctica
|
- |
|
|
534641 |
Amano Lipase PS from Pseudomonas cepacia
|
- |
|
|
62309 |
Lipase from Pseudomonas cepacia
|
- |
|
|
62279 |
Lipase, immobilized in Sol-Gel-AK from Pseudomonas cepacia
|
- |
|
|
17261 |
Lipase, immobilized on ceramic particle from Pseudomonas cepacia
|
- |
|
References:
(1) Ward, R. S. Tetrahedron: Asymmetry 1995, 6, 1475.
(2) Shvo, Y. et al. J. Am. Chem. Soc. 1986, 108, 7400.
(3) Prabhakaran, R. Synlett 2004, 2048, and ref. cit. therein.
(4) For a review, see: Pàmies, O.; Bäckvall, J.-E. Chem. Rev. 2003, 103,
3247.
|