Chemical Synthesis
Product Catalog
Technology Spotlights
ChiroSolv Kits
APhos
Chiral Phosphoric Acids
MOFs
MeOBIPHEP
PTS Amphiphile
Takasago Ligands
catASium
cataCXium
Key Resources
Drug Discovery
Materials Science
Stable Isotopes (ISOTEC)
 cataCXium® —Ligands and Complexes for Efficient Cross-Coupling Reactions
Chemical Synthesis
Introduction
Advantages
Representative Applications
Introduction
Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlorides for these transformations is preferred due to their low cost and their availability. However, despite the tremendous progress made in catalyst design, the Pd loading for these reactions
(1–3 mol%) prohibits industries to using them on a kilogram scale. The quest to synthesize a very active catalyst with very low loading has been ongoing for the past 20 years.
cataCXium® A Structure cataCXium® AHI Structure cataCXium® ABn Structure
cataCXium® A
671479
cataCXium® AHI
671584
cataCXium® ABn
671800
cataCXium® PCy Structure cataCXium® PiCy Structure cataCXium® PinCy Structure cataCXium® PintB Structure
cataCXium® PCy
672017
cataCXium® PiCy
672122
cataCXium® PinCy
672246
cataCXium® PintB
672343
cataCXium® POMeCy Structure cataCXium® POMetB Structure cataCXium® PtB Structure
cataCXium® POMeCy
672459
cataCXium® POMetB
672564
cataCXium® PtB
672661
cataCXium® FPrPh Structure cataCXium® FBu Structure cataCXium® FSulf Structure
cataCXium® FPrPh
672785
cataCXium® FBu
688681
cataCXium® FSulf
688800
cataCXium® C Structure
cataCXium® C
671916
back to top
Advantages
  • Atom economical method
  • High yields
  • High turnover numbers
Representative Applications
Suzuki Coupling with Low Loading of Catalyst
Beller and co-workers developed a very bulky ligand based on diadamantylalkylphosphanes.1,2,3 Among the most active ligand synthesized by this group, diadamantyl-n-butylphosphane, proved to be the most efficient. Using 0.005 mol% of Pd(OAc)2 and 0.01 mol% of phosphine, good to excellent yields were reported (58–100%) (Scheme 1). It is important to note that ortho substituents on the aryl chloride are tolerated and that only strongly electron donating substituents such as the methoxy group led to a drop in yields (60%) (Table 1).
Scheme 1
Scheme 1 Image
Table 1
Ar—Cl Yield TON
Table 1 Structure 1 Image 87% 17,400
Table 1 Structure 2 Image 68% 13,600
Table 1 Structure 3 Image 64% 12,800
Table 1 Structure 4 Image 100% 20,000
Table 1 Structure 5 Image 99% 19,800
back to top
Evonik in collaboration with professor Beller and co-workers, developed a novel class of phosphine ligands, the phosphino-substituted N-aryl pyrroles.4 These biaryl ligands are sterically demanding and electron rich. When applied to the reaction of a variety of aryl chlorides with phenyl boronic acid, they showed very high turnover numbers at mild reaction temperatures. Low catalyst loading (0.05 mol%) was needed for the cross-coupling of a variety of aryl chlorides with phenylboronic acid (Scheme 2). Utilizing this ligand, with Pd(OAc)2 or Pd2(dba)3, for the conversion of electron rich and electron deficient aryl chlorides, resulted in excellent yields. Table 2 summarizes the yield reported for a variety of aryl chlorides reacted with phenylboronic acid using 0.01 mol% of Pd2(dba)3 at 60 °C.
Scheme 2
Scheme 2 Image
Table 2
Ar—Cl Yield TON
Table 2 Structure 1 Image >99% 10,000
Table 2 Structure 2 Image >99% 10,000
Table 2 Structure 3 Image >99% 10,000
Table 2 Structure 4 Image 99% 9,900
back to top
Building on the success synthesizing phosphino-N-aryl pyrroles, Beller and coworkers developed a similar ligand. They synthesized phosphino-N-phenylindole based ligands.5 To assess the reactivity of these ligands, they carried out the cross-coupling reaction of chlorobenzene with aniline (Scheme 3). This new system proved to be the best among the different ligands developed by Beller and co-workers for this specific cross-coupling reaction. Using Pd(OAc)2 and the tert-butyl phosphine phenyl indole ligand with NaOtBu as a base in toluene, a wide variety of amines were arylated with excellent yields.
Scheme 3
Scheme 3 Image
References
  1. Zapf, A. et al. Angew. Chem., Int. Ed. 2000, 39, 4153.
  2. Köllhofer, A. et al. Angew. Chem., Int. Ed. 2003, 42, 1056.
  3. Ehrentraut, A. et al. Adv. Synth. Catal. 2002, 344, 209.
  4. Zapf, A. et al. Chem. Commun. 2004, 38.
  5. Rataboul, F. et al. Chem.-Eur. J. 2004, 10, 2983.
back to top
Cross-coupling Reactions
A new class of phosphorus ligand was developed by Plenio and co-workers based on fluorenylphosphines architecture.1 To test the potency of these new ligands, the team of scientists carried out several common cross-coupling reactions: Suzuki, Sonogashira and Buchwald-Hartwig amination (Figure 1). The ligand proved to be very active for the Suzuki coupling of a variety of aryl chlorides with 4-methylphenylboronic acid with loading of 0.05 to 0.5 mol% of palladium. The electron donating substituent such as OMe did not affect the yields with >99% conversion. The Sonogashira transformation was carried out with a variety of aryl chloride coupled to phenylacetylene. With 1 mol% loading of catalyst, 44% to 94% yields were reported. Finally, the Buchwald-Hartwig amination results were very promising with quantitative conversion reported for several reactions with catalyst loading as low as 0.1 mol%.
Figure 1
Figure 1 Image
References
  1. Fleckenstein, C. A.; Plenio, H. Chem.-Eur. J. 2007, 13, 2701.
back to top
Heck Catalysis
Hermann and co-workers developed a new class of palladium catalyst called palladacycles.1 These complexes are thermally stable up to 100 °C and are very potent for Heck catalysis with TON > 10,000. Tietze et al. utilized the activity of this catalyst to synthesize a derivative of cephalostatin, a steroid that showed potency against cancer cells.2 A double coupling reaction was carried out with an overall yield of 80%.
Heck Catalysis Image
References
  1. Herrmann, W. A. et al. Angew. Chem., Int. Ed. Engl. 1995, 34, 1844.
  2. Tietze, L. F.; Krahnert, W.-R. Chem. Eur. J. 2002, 8, 2116.
back to top
Product Information
Product # Product Name Structure Add to Cart
671479 cataCXium® A cataCXium® A Structure Image Shopping Cart
671584 cataCXium® AHI cataCXium® AHI Structure Image Shopping Cart
671800 cataCXium® ABn cataCXium® ABn Structure Image Shopping Cart
672017 cataCXium® PCy cataCXium® PCy Structure Image Shopping Cart
672122 cataCXium® PICy cataCXium® PICy Structure Image Shopping Cart
672246 cataCXium® PInCy cataCXium® PInCy Structure Image Shopping Cart
672343 cataCXium® PIntB cataCXium® PIntB Structure Image Shopping Cart
672459 cataCXium® POMeCy cataCXium® POMeCy Structure Image Shopping Cart
672564 cataCXium® POMetB cataCXium® POMetB Structure Image Shopping Cart
672661 cataCXium® PtB cataCXium® PtB Structure Image Shopping Cart
672785 cataCXium® FPrPh cataCXium® FPrPh Structure Image Shopping Cart
688681 cataCXium® FBu cataCXium® FBu Structure Image Shopping Cart
688800 cataCXium® FSulf cataCXium® FSulf Structure Image Shopping Cart
671916 cataCXium® C cataCXium® C Structure Image Shopping Cart
671355 Evonik Ligand Kit for CX Coupling Reactions Kit includes: 671479, 671584, 671800, 672017, 672122, 672246, 672343, 672459, 672564, 672661, 672785, 688681, 688800, 671916 Shopping Cart