Drug Discovery
Discovery CPR
Peptide Libraries
Screening Compounds
Validation Libraries
Coated Multiwell Plates
Medicinal Chemistry
Peptides
Polymer Supported Rgts
Resin Explorer
New Products
Key Resources
Product Highlights
Materials Science
Stable Isotopes (ISOTEC)
 New Products
Drug Discovery
The following product lists are available for download in PDF and SDfile formats. Both are searchable, and include structure images of every building block.
Heterocyclic Building Blocks
PDF  (571 KB)     SDfile  (172 KB)
Screening Compounds
PDF  (1.6 MB)     SDfile  (424 KB)
Polymer-Supported Reagents
Palladium Catalyst
Metal Scavengers
Reagents
Oxidizing Reagents
Base
Lewis Acids
Janda Jels™
Palladium Catalyst  
Product NameLoading Mesh Size Polymer Matrix Product # 
Palladium (II) acetate triphenylphosphine, polymer-bound 1.0-1.5 mmol Pd/g 200-400PS cross-linked with 1% DVB 654167
Bis(diphenylphosphanyl)methylamine palladium (II) acetate, polymer-bound 0.5-1.0 mmol Pd/g 70-90PS cross-linked with 1% DVB 654159
N-Methylimidazolium palladium (II), polymer-bound 0.5-1.5 mmol 50-100PS cross-linked with 1% DVB 650927
Dichlorobis(triphenylphosphine)palladium (0), polymer-bound 1.0-2.0 mmol/g 100-200PS corss-linked with 2% DVB 596930
Metal Scavengers   back to top
Product NameLoading Mesh Size Polymer Matrix Product # 
Ethylenediaminetriacetic acid acetamide, polymer-bound 3.5-4.0 mmol N/g 50-100 PS cross-linked with 1% DVB 656844
N,N,N'-Trimethylethylenediamine, polymer-bound 2.5-3.0 mmol N/g 100-200PS cross-linked with 1% DVB 656836
6-Thioicotinamide, polymer-bound 2.0-3.0 mmol/g 70-90PS cross-linked with 1% DVB 643904
Reagents   back to top
Product NameLoading Mesh Size Polymer Matrix Product # 
2-Chloro-1, 3,2-diazaphospholidine-2-sulfide, polymer-bound 1-2 mmol/g 100-200PS cross-linked with 1% DVB 657166
Methylphenylvinylsilane, polymer-bound 1.0-1.5 mmol Si/g 50-100 PS cross-linked with 1% DVB 655473
Triphenylphosphine oxide, polymer-bound2.5-3.0 mmol P/g 200-400 PS cross-linked with 2% DVB 655430
4-Hydroxyphenyl, polymer-bound 0.5-1.5 mmol OH/g 50-100PS cross-linked with 1% DVB 650382
Oxidizing Reagents   back to top
Product NameLoading Mesh Size Polymer Matrix Product # 
Iodylbenzamide, polymer-bound 0.5-1.0 mmol I/g 50-100PS cross-linked with 1% DVB 654175
Base   back to top
Product NameLoading Mesh Size Polymer Matrix Product # 
Pyridine on Rasta Resin 6.5-7.5 mmol N/g 50-100PS cross-linked with 1% DVB 655457
Lewis Acids   back to top
Product NameLoading Mesh Size Polymer Matrix Product # 
Ytterbium (II) polystyrenesulfonate, polymer-bound 0.5-1.5 mmol/g 30-80 Macroporous 641030
Scandium trifluoromethanesulfonate, polymer-bound 0.5-1.5 mmol/g 30-80Macroporous 590312
JandaJel™   back to top
Product NameLoading Mesh Size Polymer Matrix Product # 
JandaJel™-4-(methylthio)phenol 0.5-1.0 mmol S/g 50-100 PS cross-linked with 1% DVB 656879
     back to top