Drug Discovery
Discovery CPR
Peptide Libraries
Screening Compounds
Validation Libraries
Coated Multiwell Plates
Medicinal Chemistry
Peptides
Building blocks
Reagents
Resins
Unnatural Amino Acids
Polymer Supported Rgts
Resin Explorer
New Products
Key Resources
Product Highlights
Materials Science
Stable Isotopes (ISOTEC)
 BUILDING BLOCKS for PEPTIDE SYNTHESIS
Peptide and Peptidomimetic Synthesis
Sigma-Aldrich offers a comprehensive portfolio of high quality building blocks for peptide synthesis. All proteinacious amino acids are available with multiple combinations of different protecting groups and schemes — from standard building blocks for Boc-/Bzl- or Fmoc-/tBu- Solid Phase Peptide Synthesis to Z- Na-protected compounds for Solution Phase Synthesis. Orthogonal and other selectively removable protecting groups extend the scope of peptide synthesis to include unnatural, cyclic, phosphorylated, and labeled peptides.