Drug Discovery
VersaFlash™
|
A novel, quick, and inexpensive solution for flash purification of natural products and organic synthesis reaction products.
Optimize your flash chromatography with VersaFlash™, an efficient and flexible solution for discovery and development scale flash purification. The patent pending VersaFlash™ station, together with the unique VersaPak™ cartridges, represent a cost-effective and flexible system for sample separations.
- Increase recovery and purity with spherical silica cartridges
- Scale-up to larger cartridges in seconds without additional hardware
- Increase sample throughput with quick and easy cartridge changeout
- Compatible with most solvent delivery systems
- Stackable cartridges for increased purification capabilities
- Reversible cartridges for faster elution of fully-retained components
- Flexible sample loading options
Click here for detailed information on the VersaFlash™ system or take a look at Supelco's complete line of chromatography solutions.