Drug Discovery
Discovery CPR
Peptide Libraries
Screening Compounds
Validation Libraries
Coated Multiwell Plates
Medicinal Chemistry
Peptides
Polymer Supported Rgts
Resin Explorer
Solid-Phase Resins
Solution-Phase Resins
Functionalized Silica Gels
PEG Crosslinked Resins
JandaJels
Pd Scavengers/Catalysts
Peptide Resins
Phoxime Resin
Polymer Supported
Rasta Resins
New Products
Key Resources
Product Highlights
Materials Science
Stable Isotopes (ISOTEC)
 PEG CROSSLINKED RESINS
Resin Explorer
  Cl-Functionalized Resins
OH-Functionalized Resins
Soluble PEG-Based Resins
Cl-Functionalized Resins  
Product NameLoading Mesh size Product # 
Poly(ethylene glycol)(16) crosslinked Merrifield resin 0.8-1.5 mmol/g 100-200 56,488-5
Poly(ethylene glycol)(4) crosslinked Merrifield resin 1 % cross-linked with divinylbenzene 0.8-1.5 mmol/g 100-200 56,489-3
Poly(ethylene glycol)(9) crosslinked Merrifield resin 0.8-1.5 mmol/g 100-200 56,490-7
OH-Functionalized Resins
back to top
Product NameLoading Mesh size Product # 
Poly(ethylene glycol)(16) crosslinked 4-benzyloxybenzyl alcohol 0.8-1.5 mmol/g 100-200 56,482-6
Poly(ethylene glycol)(9) crosslinked 4-benzyloxybenzyl alcohol 0.8-1.5 mmol/g 100-200 56,483-4
Poly(ethylene glycol)(4) crosslinked 4-benzyloxybenzyl alcohol 0.8-1.5 mmol/g 100-200 56,484-2
Soluble PEG-Based Resins
back to top
Product NameProduct # 
Poly(ethylene glycol) triphenylphosphine 53,264-9
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt bound to PEG 750 54,323-3