Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Drug Discovery > Resin Explorer > Phoxime Resin
Resin Explorer
Phoxime Resin
Phoxime resin is a novel, air-stable, easily handled polymer-bound phosgene equivalent. Unlike typical chloroformates, little to no decomposition was observed on prolonged exposure to air.6 Heat-labile polymer-bound oxime carbamate intermediates synthesized from the addition of isocyanates to Phoxime resin have been shown to liberate latent isocyanates upon thermolysis. The subsequent addition of amines to solution affords ureas in excellent yields and high chemical purity.6 The graph below illustrates the temperature dependence on the urea formation.7
Catalog No. Description Structure
52,373-9 Phoxime Resin 523739
Chart
First International Electronic Conference on Synthetic Organic Chemistry (ECSOC-1),
www.unibas.ch/mdpi/ecsoc/b0001.htm,
September 1-30, 1997
  1. Lam, K.S. et. al. Tetrahedron Lett. 1997, 97, 411.
  2. Scialdone, M.A., J. Org. Chem. 1998, 63, 4802.
  3. DeGrado, W.F.; Kaiser, E.T. J. Org. Lett. 1980, 45, 1295.
  4. DeGrado, W.F.; Kaiser, E.T. J. Org. Lett. 1982, 47, 3258.
  5. Scialdone, M.A., Tetrahedron Lett. 1996, 37, 8141.
  6. Scialdone, M.A., J. Org. Chem. 1998, 63, 4802.
  7. Scialdone, M.A., J. Org. Chem. 1998, 63, 4802.
  8. Scialdone, M.A., J. Org. Chem. 1998, 63, 4802
  9. MacDonald, A.A. et al. First International Electronic Conference on Synthetic Organic Chemistry (ECSOC-1), www.unibas.ch/mdpi/ecsoc/b0001.htm September 1-30, 1997