Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Drug Discovery > Resin Explorer > Resins for Solid-Phase Synthesis
Resin Explorer
Resins for Solid-Phase Synthesis
  Alkenyl Resins
Amine-Functionalized Resins
BHA Resins
Br-Functionalized Resins
Chloromethyl Resins
CHO-Functionalized Resins
Cl-Functionalized Resins
COOH-Functionalized Resins
HypoGel® Resins
I-Functionalized Resins
JandaJels™
MBHA Resins
Miscellaneous Resins
NH2-Functionalized Resins
Nitrophenyl Carbonates
OH-Functionalized Resins
Oxime Resins
Peg Cross-Linked
Boc-/Bzl Peptide Synthesis
Fmoc-/tBu Peptide Synthesis
Phoxime Resin
Resin Matrices
Rink Acid Resins
Rink Resins
Safety Catch Resins
TentaGel® Resins
Thiol-Functionalized Resins
Triazine-Based Resins
Trityl Amines
Wang Resins
Wang Derivatives
Alkenyl Resins        
Product Name Loading Mesh Size Polymer Matrix Product # 
REM resin 0.1-0.5 mmol/g 200-400 PS crosslinked with 1% acrylate ester 471739
REM resin ca.1.0 mmol/g 200-400 PS crosslinked with 1% DVB 510173
REM resin 1.1 mmol/g 100-200 PS crosslinked with 1% DVB 537608
REM resin ca. 0.5 mmol/g 70-90 PS crosslinked with 1 % DVB 537381
REM resin ~ 0.9 mmol/g 100-300 PS crosslinked with 1 % DVB 83558
Vinyl sulfone, polymer-bound 0.4-1.2 mmol/g 100-200 PS crosslinked with 1% DVB 564737
Vinyl-polystyrene Capacity ~1.5 mmol/g, ~ 1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 17761
Amine-Functionalized Resins        
Product Name Loading Mesh Size Polymer Matrix Product # 
Amidine resin ~ 0.9 mmol/g 100-300 PS crosslinked with 2% DVB 06565
N-(4-Benzyloxybenzyl)hydroxylamine polymer bound Capacity ~0.5 mmol/g, resin ~ 0.5 mmol/g 200-400 PS crosslinked with 1 % DVB 17762
(Aminomethyl)polystyrene 1.0-1.5 mmol/g 200-400 PS crosslinked with 1% DVB 474045
(Aminomethyl)polystyrene 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 514772
(Aminomethyl)polystyrene 1.0 mmol/g 100-200 PS crosslinked with 1% DVB 522759
(Aminomethyl)polystyrene 0.5-1.0 mmol/g 70-90 PS crosslinked with 1% DVB 515612
(Aminomethyl)polystyrene Particle size 70-90 mesh 1.0-1.5 mmol/g 70-90 PS crosslinked with 1% DVB 515620
(Aminomethyl)polystyrene 1.5-2.0 mmol/g 70-90 PS crosslinked with 1% DVB 515639
(R)-(+)-a-methylbenzylamine, polymer-bound ca.2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 576395
2-Chlorotrityl Knorr resin 0.5-1.0 mmol/g 100-200 PS crosslinked with 1% DVB 549401
2-N-Fmoc-Amino-dibenzocyclohepta-1,4-diene, polymer-bound 0.5-1.3 mmol/g 100-200 PS crosslinked with 1% DVB 564672
4-[4-(1-Fmoc-aminoethyl)-2-methoxy-5-nitrophenoxy]butyramidomethyl-polystyrene 0.6-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564753
4-Benzyloxybenzylamine, polymer-bound  0.5-1.3 mmol/g 100-200 PS crosslinked with 1 % DVB 564818
4-Carboxybenzenesulfonamide, polymer-bound 0.8-1.0 50-90 PS crosslinked with 1% DVB 534625
4-Carboxybenzenesulfonamide, polymer-bound 0.7-1.3 mmol/g 100-200 PS crosslinked with 1% DVB 549347
(Aminomethyl)polystyrene macroporous 1.5-3.0 mmol/g 30-60 PS crosslinked with 1% DVB 564095
(Aminomethyl)polystyrene macroporous 1.5-3.0 mmol/g 70-90 PS crosslinked with 1% DVB 564109
Bis(tert-butoxycarbonyl)thiopseudourea, polymer-bound ca. 1.0 mmol/g 100-200 PS crosslinked with 1% DVB 540382
Dimethylaminomethyl-polystyrene 1.0-1.5 mmol/g 20-50 PS crosslinked with 1% DVB 426997
Fmoc-3-amino-3-(2-nitrophenyl)propionic acid, polymer-bound 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564745
N-Methyl aminomethylated polystyrene 1.0-1.5 mmol/g 100-200 PS crosslinked with 1% DVB 538779
PAL resin 0.4-0.8 mmol/g 100-200 PS crosslinked with 1% DVB 549444
Sieber amide resin 0.3-0.6 mmol/g 50-100 PS crosslinked with 1% DVB 533955
tert-Butyl N-(2-mercaptoethyl)carbamate, polymer-bound ca. 2 mmol/g 100-200 PS crosslinked with 1% DVB 579378
Triphenylchloromethane-4-carboxamide polymer bound 1.1-1.3 mmol/g 200-400 PS cross-linked with 1% DVB 00481
BHA Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
2-Chlorobenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572632
2-Chlorobenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572632
HMPB-benzhydrylamine polymer bound 0.7-1.3 mmol/g 100-200 PS cross-linked with 1% DVB 564850
HMPB-benzhydrylamine polymer bound 0.7-1.3 mmol/g 100-200 PS cross-linked with 1% DVB 564850
4-Methylbenzhydrol, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 564206
4-Methylbenzhydrol, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 564206
4-Methylbenzhydryl alcohol, polymer-bound 1.6-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 572624
4-Methylbenzhydryl alcohol, polymer-bound 1.6-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 572624
4-Methylbenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572640
4-Methylbenzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572640
Benzhydrol, polymer-bound 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 564192
Benzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572616
Benzhydryl chloride, polymer-bound 1.6-2.2 mmol/g 100-200 PS cross-linked with 1% DVB 572616
Benzhydrylamine polymer-bound hydrochloride 0.7-1.4 mmol/g 100-200 PS cross-linked with 1% DVB 532460
Benzhydrylamine polymer-bound hydrochloride 0.7-1.4 mmol/g 200-400 PS cross-linked with 1% DVB 532479
Benzhydrylamine polymer-bound hydrochloride 0.7-1.4 mmol/g 100-200 PS cross-linked with 2% DVB 532436
Benzhydrylamine hydrochloride polymer bound ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 12106
Benzhydrylamine hydrochloride polymer bound ~ 1.5 mmol/g 100-200 PS cross-linked with 2% DVB 12110
Br-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-(Benzyloxy)benzyl bromide polymer bound 0.5-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 534854
4-Bromopolystyrene 0.5-2.0 mmol/g 100-200 PS cross-linked with 1% DVB 549355
Brominated PPOA resin 0.5-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 533920
Brominated Wang resin 0.5-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 534471
Brominated Wang resin ~ 1.0 mmol/g 70-90 PS cross-linked with 1% DVB 10984
Bromoacetal, polymer-bound 1.0-1.5 mmol/g 75-100 PS cross-linked with 1% DVB 564869
Bromoacetic acid, polymer-bound 0.85-1.2 mmol/g 100-200 PS cross-linked with 1% DVB 537500
Bromopolystyrene 1.2-1.3 mmol/g 100-400 PS cross-linked with 2% DVB 18106
Bromopolystyrene ~ 2.5 mmol/g 100-400 PS cross-linked with 2% DVB 18099
Bromopolystyrene ~4 mmol/g 200-400 PS cross-linked with 1% DVB 18100
Bromopolystyrene ~ 4 mmol/g 200-400 PS cross-linked with 2% DVB 18104
Bromopolystyrene ~ 5.5 mmol/g 200-400 PS cross-linked with 2% DVB 18102
Bromopolystyrene ~ 5.5 mmol/g 100-400 PS cross-linked with 2% DVB 18101
HypoGel® 200 Br ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 79709
Polystyrene A-Br for peptide synthesis 0.8-1.2 mmol/g 160-200 mm PS cross-linked with 1% DVB 81534
Polystyrene A-Br for peptide synthesis 0.8-1.2 mmol/g 500-560 mm PS cross-linked with 1% DVB 81535
Selenium bromide, polymer-bound       535958
TentaGel HL-Br ~ 0.48 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86331
TentaGel MB-Br ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86353
TentaGel S-Br ~ 0.26 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86363
TentaGel S-Br ~ 0.25 mmol/g ~ 130 mm PEG grafted on low cross-linked PS beads 86357
Chloromethyl Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
[5-[4-(Chloromethyl)phenyl]pentyl]styrene, polymer-bound 0.75-1.25 mmol/g 100-200 PS cross-linked with 1% DVB 513776
4-(Benzyloxy)benzyl chloride polymer bound 0.5-1.0 mmol/g 100-200 PS cross-linked with 1% DVB 537276
4-Methoxybenzhydryl chloride, polymer-bound ca.1.0 mmol/g 50-90 PS cross-linked with 1% DVB 534692
CHO-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
(4-Formyl-3-methoxyphenoxymethyl)polystyrene 1.0-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 516449
(4-Formyl-3-methoxyphenoxymethyl)polystyrene ~ 1.2 mmol/g 100-300 PS crosslinked with 2% DVB 09942
3-Benzyloxybenzaldehyde, polymer-bound 1.5-2.0 mmol/g 200-400 PS cross-linked with 1% DVB 477885
4-(4-Formyl-3-methoxyphenoxy)butyryl AM resin 0.8-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 541583
4-Benzyloxy-2,6-dimethoxybenzaldehyde,polymer-bound 1.0-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 538213
Formylpolystyrene 0.7-1.5 mmol/g 100-200 PS cross-linked with 1% DVB 532401
Formylpolystyrene 2.0-3.0 mmol/g 100-200 PS cross-linked with 2% DVB 532428
HypoGel® 200 CHO ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 75401
HypoGel® 200 FP ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB;) PS beads 78528
Indole resin ca. 0.5 mmol/g 200-400 PS crosslinked with 1 % DVB 516279
Indole resin ~ 1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 20529
Polystyrene A-CH(OEt)2 0.8-1.2 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81537
Polystyrene A-CH(OEt)2 0.8-1.2 mmol/g 500-560 mm PS crosslinked with 1 % DVB 81538
Polystyrene AM-CH(OEt)2 Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 81543
Polystyrene AM-CH(OEt)2 for peptide synthesis Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 200-400 PS crosslinked with 1 % DVB 81544
Polystyrene AM-CH(OEt)2 Capacity (N) 0.8-1.2 mmol/g, resin 0.8-1.2 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81545
TentaGel HL-CH(OEt)2 Capacity ~0.40 mmol/g, ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86332
TentaGel MB-CH(OEt)2 Capacity ~0.40 mmol/g, ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86354
Cl-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Benzoyl chloride polymer bound ~ 2.1 mmol/g 100-300 PS crosslinked with 2% DVB 12973
(Chloromethyl)polystyrene ~ 0.8 mmol/g 200-400 PS cross-linked with 1% DVB 63865
(Chloromethyl)polystyrene ~ 1.1 mmol/g 200-400 PS cross-linked with 1% DVB 63852
(Chloromethyl)polystyrene ~ 1.7 mmol/g 200-400 PS cross-linked with 1% DVB 63866
(Chloromethyl)polystyrene ~ 4.5 mmol/g 200-400 PS cross-linked with 1% DVB 63863
(Chloromethyl)polystyrene ~ 1.5 mmol/g 70-90 PS cross-linked with 1% DVB 10983
(Chloromethyl)polystyrene ~ 2 mmol/g 250-300 mm PS cross-linked with 1% DVB 17296
(Chloromethyl)polystyrene ~ 1.0 mmol/g 400-500 mm PS cross-linked with 1% DVB 17298
(Chloromethyl)polystyrene ~ 2 mmol/g 400-500 mm PS cross-linked with 1% DVB 17297
(Chloromethyl)polystyrene gel type ~ 2 mmol/g 70-150 PS cross-linked with 1.5% DVB 63858
(Chloromethyl)polystyrene ~ 1.4 mmol/g 200-400 PS cross-linked with 2% DVB 63872
(Chloromethyl)polystyrene ~ 2.1 mmol/g 200-400 PS cross-linked with 2% DVB 63874
(Chloromethyl)polystyrene >= 4.0 mmol/g 100-200 PS crosslinked with 2% DVB 75063
(Chloromethyl)polystyrene ~ 4.3 mmol/g 200-400 PS cross-linked with 2% DVB 63873
(Chloromethyl)polystyrene gel type ~ 2 mmol/g 70-150 PS cross-linked with 2.5% DVB 63861
(Chloromethyl)polystyrene gel type ~ 2 mmol/g 18-70 PS cross-linked with 2.5% DVB 63862
(Chloromethyl)polystyrene gel type ~ 5 mmol/g 16-50 PS cross-linked with 3.5% DVB 63864
(Chloromethyl)polystyrene gel type ~ 5 mmol/g 16-50 PS cross-linked with 4.5% DVB 63867
(Chloromethyl)polystyrene ~ 5.5 mmol/g 16-50 PS cross-linked with 5.5% DVB 63868
(Chloromethyl)polystyrene gel type ~ 5.5 mmol/g 16-50 PS cross-linked with 5.8% DVB 63875
(Chloromethyl)polystyrene ~ 0.7 mmol/g 200-400 PS cross-linked with 2% DVB 63871
Chloroacetyl, polymer-bound 2.5-5.0 mmol/g 100-200 PS crosslinked with 1 % DVB 564915
Merrifield's resin 2.5-4.0 mmol/g 50-100 PS crosslinked with 1 % DVB 577901
Merrifield's resin 1.0-1.5 mmol/g 200-400 PS crosslinked with 1 % DVB 221481
Merrifield's resin 1.0-1.5 mmol/g 200-400 PS crosslinked with 2 % DVB 221503
Merrifield's resin 1.5-2.0 mmol/g 200-400 PS crosslinked with 1 % DVB 437956
Merrifield's resin 2.0-2.5 mmol/g 200-400 PS crosslinked with 2 % DVB 449113
Merrifield's resin 3.0-3.5 mmol/g 200-400 PS crosslinked with 1 % DVB 456969
Merrifield's resin 3.5-4.5 mmol/g 200-400 PS crosslinked with 1 % DVB 474517
Merrifield's resin 0.5-1.0 mmol/g 100-200 PS crosslinked with 1 % DVB 516074
Merrifield's resin 0.8-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 522716
Merrifield's resin 1.0-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 515957
Merrifield's resin 1.5-2.0 mmol/g 100-200 PS crosslinked with 1 % DVB 497045
Merrifield's resin 2.0-3.0 mmol/g 100-200 PS crosslinked with 1 % DVB 49,705-3
Merrifield's resin 3.5-4.5 mmol/g 100-200 PS crosslinked with 1 % DVB 49,706-1
Merrifield's resin 1.0-1.5 mmol/g 70-90 PS crosslinked with 1 % DVB 49,919-6
Merrifield's resin 1.5-2.0 mmol/g 70-90 PS crosslinked with 1 % DVB 49,918-8
Merrifield's resin 1.7-2.2 mmol/g 50-100 PS crosslinked with 1 % DVB 52,272-4
Merrifield's resin 1.7-2.2 mmol/g 50-100 PS crosslinked with 1 % DVB 52,273-2
Merrifield's resin 3.6-4.2 mmol/g 50-100 PS crosslinked with 1 % DVB 52,274-0
COOH Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
Carboxyethylpolystryrene 0.8-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 53,862-0
Carboxypolystyrene 0.5-1.5 mmol/g 100-200 PS crosslinked with 1 % DVB 53,237-1
Carboxypolystyrene 1.6-3.0 mmol/g 100-200 PS crosslinked with 1 % DVB 53,239-8
HypoGel® 200 COOH ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 78967
Polystyrene A-COOH 0.8-1.2 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81536
Polystyrene A-COOH 0.8-1.2 mmol/g 500-560 mm PS crosslinked with 1 % DVB 81539
Polystyrene AM-COOH 0.8-1.2 mmol/g 100-200 PS crosslinked with 1 % DVB 81546
Polystyrene AM-COOH for peptide synthesis 0.8-1.2 mmol/g 200-400 PS crosslinked with 1 % DVB 81547
Polystyrene AM-COOH 0.6-0.8 mmol/g 160-200 mm PS crosslinked with 1 % DVB 81548
TentaGel HL-COOH ~ 0.40 mmol/g ~ 110 mm PEG grafted on low cross-linked PS beads 86333
TentaGel MB-COOH ~ 0.40 mmol/g 140-170 mm PEG grafted on low cross-linked PS beads 86355
TentaGel S-COOH ~ 0.25 mmol/g ~ 90 mm PEG grafted on low cross-linked PS beads 86336
TentaGel S-COOH ~ 0.25 mmol/g ~ 130 mm PEG grafted on low cross-linked PS beads 86358
HypoGel® Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
HypoGel® 200 FMP ~ 0.6 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 74911
HypoGel® 200 PHB ~ 0.7 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 75513
HypoGel® 200 Trt-OH ~ 0.7 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 75040
HypoGel® 200 HMB ~ 0.8 mmol/g 110-150 mm PEG (n = 5) grafted on low cross-linked (1% DVB) PS beads 78619
I-Functionalized Resins       back to top
Product Name Loading Mesh Size Polymer Matrix Product # 
4-Iodophenol, polymer-bound 1.0-2.0 mmol/g 50-100 PS crosslinked with 1 % DVB 570788
Iodopolystyrene 3.5-4.0 mmol/g 50-100