New Products
Nanotechnology
Biocompatible/Biodegradable
Biocompatible Inorganics
Polymers
Polyphosphoesters
Tutorial
Display and Optoelectronics
Energy Source Materials
Metals and Ceramic Science
Micro and Nanoelectronics
Nanomaterials
Polymer Science
Key Resources
Stable Isotopes (ISOTEC)
 POLYPHOSPHOESTERS
Biocompatible and Biodegradable
Polyphosphoesters exhibit many properties necessary for drug delivery and gene delivery.
  • Structurally similar to nucleic and teichoic acids.
  • Biodegrade through hydrolysis and possibly enzymatic digestion.
  • One of the few classes of polymers for non-viral gene delivery by conjugation of charged groups to the side chain of the phosphate.
659606   Poly((lactide-co-ethyleneglycol)-co-ethyloxyphosphate)
Poly(LAEG-EOP)
Solubility:
THF:    soluble (readily)
Chloroform:    soluble (readily)
Acetonitrile:   soluble (readily)
Ethyl Acetate:  soluble (readily)
Hexane:    insoluble
Change in molecular weight as a function of time, during the in vitro and in vivo degradation of Poly(LAEG-EOP).1
659614   Poly(1,4-bis(hydroxyethyl)terephthalate-co-ethyloxyphosphate)
Poly(BHET-EOP)
Solubility:
THF:     insoluble
Hexane:   insoluble
Chloroform:   soluble (readily)
Methylene chloride:  soluble (readily)
Water:    swells (opaque solution)
659738  Poly(1,4-bis(hydroxyethyl)terephthalate-alt-ethyloxyphosphate)-co-1,4-bis(hydroxyethyl)terephthalate-co-terephthalate)
Poly(BHET-EOP/TC, 80/20)
Solubility:
THF:     swells (insoluble)
Hexane:   swells (insoluble)
Chloroform:  soluble
Reference
1)  Zhan, Z. et. al. Adv. Drug Delv. Rev. 2003,55, 483
See also: Penczek, S. et al. Polish J. Chem., 2001, 75, 1171. Penczek, S. et al. Polish J. Chem., 1994, 68, 2129