Sigma-Aldrich
Spacer
Materials Science > Organic Electronics
Materials Science
Organic Electronics
The organic electronics and photonics product line consists of three groups: organic electronics, photonics, and liquid crystals. Organic electronics materials are used to make organic light-emitting displays (OLEDs), field-effect transistors (OFETs), and organic photovoltaics (OPVs). Light-emitting polymers (LEPs) showing promise for polymer light-emitting displays (PLEDs) are also available. Photonics includes a comprehensive selection of dyes and organic non-linear optical (NLO) and waveguide materials. A selection of liquid crystals and a broad range of organic electronics building blocks are also available for your research.
Products:
Technology Spotlights:
Learning Center:
Material Matters™ - Vol. 2 No. 3.  Organic Electronics.
Vol. 2, No. 3
(2.0 MB PDF)
Organic Electronics
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Aldrich Substructure Search
Subscribe to Material Matters™ Banner
Request 2008-2010 Materials Science Catalog
Coming Soon - the NEW 2009-2010 Aldrich Handbook