|
Sigma-Aldrich is pleased to offer a wide selection of ultra-high-purity, ultra-dry inorganic halides for the growth of crystals for scintillation, optics, and dosimetry. Our manufacturing facility in Urbana, Illinois has a proven track record for supplying crystal growers with ultra-high-purity source materials. Products are available from development to commercial quantities (gram to ton scale). Custom particle sizes are available upon request. For more information on our products, capabilities and services, or for a competitive quote or sample, email matsci@sial.com or call (414) 438-3831.
| Z |
Formula |
Material |
Purity |
Morphology |
Particle Size* |
Product # |
| 3 |
LiBr |
Lithium bromide |
5N |
beads |
-10 mesh |
429465 |
| 3 |
LiBr |
Lithium bromide |
4N5+ |
powder |
- |
229733 |
| 3 |
LiBr |
Lithium bromide |
3N+ |
beads |
-10 mesh |
449873 |
| 3 |
LiCl |
Lithium chloride |
4N8 |
beads |
-10 mesh |
429457 |
| 3 |
LiCl |
Lithium chloride |
3N+ |
beads |
-10 mesh |
449881 |
| 3 |
LiF |
Lithium fluoride |
4N+ |
powder |
- |
449903 |
| 3 |
LiI |
Lithium iodide |
5N |
beads |
-10 mesh |
450952 |
| 3 |
LiI |
Lithium iodide |
4N |
beads |
-10 mesh |
439746 |
| 11 |
NaBr |
Sodium bromide |
5N |
beads |
-10 mesh |
460591 |
| 11 |
NaBr |
Sodium bromide |
4N+ |
powder |
- |
229881 |
| 11 |
NaCl |
Sodium chloride |
5N |
beads |
-10 mesh |
450006 |
| 11 |
NaCl |
Sodium chloride |
5N |
powder |
- |
204439 |
| 11 |
NaCl |
Sodium chloride |
4N |
beads |
-10 mesh |
450014 |
| 11 |
NaF |
Sodium fluoride |
4N |
powder |
- |
450022 |
| 11 |
NaI |
Sodium iodide |
5N |
beads |
-10 mesh |
439681 |
| 11 |
NaI |
Sodium iodide |
5N |
powder |
- |
409286 |
| 12 |
MgCl2 |
Magnesium chloride |
4N |
beads |
-10 mesh |
449172 |
| 12 |
MgCl2 |
Magnesium chloride |
3N |
beads |
-10 mesh |
449164 |
| 12 |
MgI2 |
Magnesium iodide |
4N8 |
beads |
-10 mesh |
449911 |
| 13 |
AlBr3 |
Aluminum bromide |
5N |
powder |
- |
449601 |
| 13 |
AlCl3 |
Aluminum chloride |
5N |
powder |
- |
563919 |
| 13 |
AlCl3 |
Aluminum chloride |
4N |
powder |
- |
449598 |
| 13 |
AlF3 |
Aluminum fluoride |
3N+ |
powder |
- |
449628 |
| 19 |
KBr |
Potassium bromide |
5N |
powder |
- |
449962 |
| 19 |
KBr |
Potassium bromide |
4N |
powder |
- |
229792 |
| 19 |
KBr |
Potassium bromide |
3N5 |
powder |
- |
451010 |
| 19 |
KBr |
Potassium bromide |
3N+ |
beads |
-10
mesh |
449970 |
| 19 |
KCl |
Potassium chloride |
5N |
beads |
-10
mesh |
449989 |
| 19 |
KCl |
Potassium chloride |
5N |
powder |
- |
409316 |
| 19 |
KCl |
Potassium chloride |
4N+ |
powder |
- |
204099 |
| 19 |
KCl |
Potassium chloride |
4N |
beads |
-10 mesh |
451029 |
| 19 |
KF |
Potassium fluoride |
4N+ |
powder |
- |
449148 |
| 19 |
KF |
Potassium fluoride |
4N |
powder |
- |
229814 |
| 19 |
KI |
Potassium iodide |
4N8 |
beads |
-10 mesh |
429422 |
| 19 |
KI |
Potassium iodide |
4N+ |
powder |
- |
204102 |
| 20 |
CaBr2 |
Calcium bromide |
3N8 |
beads |
-10 mesh |
413631 |
| 20 |
CaCl2 |
Calcium chloride |
4N+ |
beads |
-10 mesh |
429759 |
| 20 |
CaCl2 |
Calcium chloride |
3N+ |
beads |
-10 mesh |
449709 |
| 20 |
CaF2 |
Calcium fluoride |
4N |
powder |
- |
449717 |
| 20 |
CaI2 |
Calcium iodide |
5N |
beads |
-10 mesh |
439797 |
| 20 |
CaI2 |
Calcium iodide |
3N5 |
beads |
-10 mesh |
516244 |
| 21 |
ScCl3 |
Scandium chloride |
4N |
powder |
- |
451266 |
| 21 |
ScCl3 |
Scandium chloride |
3N |
powder |
- |
409359 |
| 21 |
ScF3 |
Scandium fluoride |
4N |
powder |
- |
432105 |
| 24 |
CrCl2 |
Chromium(II) chloride |
4N |
powder |
- |
450782 |
| 24 |
CrCl3 |
Chromium(III) chloride |
4N |
flakes |
- |
450790 |
| 24 |
CrF3 |
Chromium(III) fluoride |
4N |
powder |
- |
449768 |
| 25 |
MnCl2 |
Manganese(II) chloride |
5N |
beads |
-10 mesh |
450995 |
| 25 |
MnCl2 |
Manganese(II) chloride |
4N |
beads |
-10 mesh |
429449 |
| 26 |
FeBr2 |
Iron(II) bromide |
5N |
beads |
-10 mesh |
434000 |
| 26 |
FeCl3 |
Iron(II) chloride |
4N8 |
beads |
-10 mesh |
429368 |
| 26 |
FeCl2 |
Iron(II) chloride |
4N |
beads |
-10 mesh |
450936 |
| 26 |
FeCl2 |
Iron(II) chloride |
3N |
beads |
-10 mesh |
450944 |
| 26 |
FeI2 |
Iron(II) iodide |
4N+ |
beads |
-10 mesh |
400858 |
| 27 |
CoBr2 |
Cobalt(II) bromide |
4N |
beads |
-10 mesh |
427136 |
| 27 |
CoCl2 |
Cobalt(II) chloride |
5N |
beads |
-10 mesh |
409332 |
| 27 |
CoCl2 |
Cobalt(II) chloride |
3N |
beads |
-10 mesh |
449776 |
| 27 |
CoF2 |
Cobalt(II) fluoride |
4N |
powder |
- |
476765 |
| 27 |
CoI2 |
Cobalt(II) iodide |
5N |
beads |
-10 mesh |
429740 |
| 29 |
CuBr2 |
Copper(II) bromide |
5N |
powder |
- |
437867 |
| 29 |
CuCl2 |
Copper(II) chloride |
5N |
powder |
- |
203149 |
| 29 |
CuCl2 |
Copper(II) chloride |
4N5+ |
powder |
- |
451665 |
| 30 |
ZnBr2 |
Zinc bromide |
5N |
beads |
-10 mesh |
451398 |
| 30 |
ZnBr2 |
Zinc bromide |
5N |
powder |
- |
230022 |
| 30 |
ZnCl2 |
Zinc chloride |
5N |
beads |
-10 mesh |
456845 |
| 30 |
ZnCl2 |
Zinc chloride |
5N |
powder |
- |
229997 |
| 30 |
ZnCl2 |
Zinc chloride |
4N5+ |
powder |
- |
429430 |
| 30 |
ZnCl2 |
Zinc chloride |
4N |
beads |
-10 mesh |
450111 |
| 30 |
ZnI2 |
Zinc iodide |
5N |
beads |
-10 mesh |
409278 |
| 30 |
ZnI2 |
Zinc iodide |
5N |
powder |
- |
466360 |
| 30 |
ZnI2 |
Zinc iodide |
4N+ |
powder |
- |
230014 |
| 31 |
GaI3 |
Gallium (III) iodide |
5N |
beads |
- |
595497 |
| 31 |
GaCl3 |
Gallium(III) chloride |
5N+ |
beads |
-10 mesh |
427128 |
| 31 |
GaCl3 |
Gallium(III) chloride |
4N |
beads |
-10 mesh |
450898 |
| 31 |
GaF3 |
Gallium(III) fluoride |
4N+ |
powder |
- |
451630 |
| 38 |
SrBr2 |
Strontium bromide |
4N5 |
beads |
-10 mesh |
566276 |
| 38 |
SrBr2 |
Strontium bromide |
4N5 |
powder |
- |
430684 |
| 38 |
SrCl2 |
Strontium chloride |
4N5 |
beads |
-10 mesh |
451282 |
| 38 |
SrCl2 |
Strontium chloride |
4N+ |
powder |
- |
439665 |
| 38 |
SrF2 |
Strontium fluoride |
4N |
powder |
- |
450030 |
| 38 |
SrI2 |
Strontium iodide |
4N+ |
beads |
-10 mesh |
400696 |
| 38 |
SrI2 |
Strontium iodide |
4N+ |
powder |
- |
466336 |
| 39 |
YBr3 |
Yttrium bromide |
3N |
beads |
-10 mesh |
450081 |
| 39 |
YBr3 |
Yttrium bromide |
3N |
powder |
- |
451355 |
| 39 |
YCl3 |
Yttrium chloride |
4N |
beads |
-10 mesh |
450103 |
| 39 |
YCl3 |
Yttrium chloride |
4N |
powder |
- |
451363 |
| 39 |
YF3 |
Yttrium fluoride |
4N |
powder |
- |
451371 |
| 40 |
ZrCl4 |
Zirconium(IV) chloride |
4N |
powder |
- |
647640 |
| 40 |
ZrF4 |
Zirconium(IV) fluoride |
4N |
powder |
- |
561460 |
| 47 |
AgCl |
Silver chloride |
5N |
powder |
- |
204382 |
| 47 |
AgCl |
Silver chloride |
4N8 |
beads |
-10 mesh |
449571 |
| 47 |
AgI |
Silver iodide |
5N |
powder |
- |
204404 |
| 48 |
CdBr2 |
Cadmium bromide |
5N |
beads |
-10 mesh |
413623 |
| 48 |
CdCl2 |
Cadmium chloride |
5N |
beads |
-10 mesh |
439800 |
| 48 |
CdCl2 |
Cadmium chloride |
4N+ |
powder |
- |
202908 |
| 48 |
CdCl2 |
Cadmium chloride |
3N+ |
beads |
-10 mesh |
449687 |
| 48 |
CdI2 |
Cadmium iodide |
5N |
beads |
-10 mesh |
449695 |
| 48 |
CdI2 |
Cadmium iodide |
5N |
powder |
- |
202916 |
| 49 |
InBr |
Indium(I) bromide |
5N |
beads |
-10 mesh |
460605 |
| 49 |
InI |
Indium(I) iodide |
5N |
beads |
-10 mesh |
413658 |
| 49 |
InCl3 |
Indium(III) chloride |
5N+ |
powder |
- |
429414 |
| 49 |
InCl3 |
Indium(III) chloride |
5N |
powder |
- |
203440 |
| 50 |
SnCl2 |
Tin(II) chloride |
4N+ |
powder |
- |
204722 |
| 50 |
SnI2 |
Tin(II) iodide |
5N |
beads |
-10 mesh |
409308 |
| 52 |
TeBr4 |
Tellurium tetrabromide |
4N5 |
beads |
-10 mesh |
451576 |
| 55 |
CsBr |
Cesium bromide |
5N |
beads |
-10 mesh |
429392 |
| 55 |
CsBr |
Cesium bromide |
5N |
powder |
- |
203017 |
| 55 |
CsBr |
Cesium bromide |
3N |
beads |
-10 mesh |
450766 |
| 55 |
CsBr |
Cesium bromide |
3N |
powder |
- |
202142 |
| 55 |
CsCl |
Cesium chloride |
5N+ |
powder |
- |
203025 |
| 55 |
CsCl |
Cesium chloride |
5N |
beads |
-10 mesh |
449733 |
| 55 |
CsCl |
Cesium chloride |
4N |
beads |
-10 mesh |
450774 |
| 55 |
CsCl |
Cesium chloride |
3N |
beads |
-10 mesh |
449741 |
| 55 |
CsF |
Cesium fluoride |
4N |
powder |
- |
255718 |
| 55 |
CsI |
Cesium iodide |
5N |
beads |
-10 mesh |
429384 |
| 55 |
CsI |
Cesium iodide |
5N |
powder |
- |
203033 |
| 55 |
CsI |
Cesium iodide |
3N |
powder |
- |
202134 |
| 56 |
BaBr2 |
Barium bromide |
5N |
beads |
-10 mesh |
413607 |
| 56 |
BaCl2 |
Barium chloride |
5N |
beads |
-10 mesh< | |