New Products
Nanotechnology
Biocompatible/Biodegradable
Energy Source Materials
Metals and Ceramic Science
Micro and Nanoelectronics
Nanomaterials
Organic Electronics
Crystal Grade Inorganics
Crystal Grade Materials
Liquid Crystals
OFET and OPV
OLED and PLED
Photonic Materials
Plexcore
Product Catalog
Substrates and Electrodes
Synthetic Tools
Polymer Science
Key Resources
Stable Isotopes (ISOTEC)
 INDIUM TIN OXIDE COATED SLIDES
Display and Optoelectronics
Prod. # Substrate Type Surface Resistance
(sq W)
Dimensions Passivation Layer
(Å, SiO2)
ITO Thickness  (Å) Optical Transmittance (%)
57,827-4 Glass slide1 8-12 25 X 75 X 1.1 mm 200-300 1200-1600 >83
63,691-6 Glass slide1 15-25 25 X 75 X 1.1 mm 200-300 600-1000 >78
63,690-8 Glass slide1 30-60 25 X 75 X 1.1 mm 200-300 300-600 >84
57,635-2 Glass slide1 70-100 25 X 75 X 1.1 mm 200-300 150-300 >87
57,636-0 Aluminosilicate2 glass slide 5-15 25 X 75 X 1.1 mm none 1200-1600 >85
63,693-2 PET3 slide 8-12 150 X 150 X 0.2 mm none 1200-1600 >84
63,692-4 PET3 slide 60-100 150 X 150 X 0.2 mm none 150-300 >76
63,931-1 PET3 sheet 40-45 1 ft X 1 ft X 5 mil none 1200 >86
63,930-3 PET3 sheet 50-70 1 ft X 1 ft X 5 mil none 1000 >79
63,928-1 PET3 sheet 90-110 1 ft X 1 ft X 5 mil none 750 >77
  1. Typical composition of slide is 72.6% SiO2, 0.8% B2O3, 1.7% Al2O3, 4.6% CaO, 3.6% MgO, and 15.2% Na2O
  2. Typical composition of slide is 55.0% SiO2, 7.0% B2O3, 10.4% Al2O3, 21.0% CaO, and 1.0% Na2O
  3. PET = poly(ethylene terephthalate)
  4. 5 mil = 0.125 mm