New Products
Nanotechnology
Alternative Energy Materials
Polymeric Electrolytes
Alloys
Electrode Materials
MOFs
Storage Applications
Tutorial
Biocompatible/Biodegradable
Metal and Ceramic Science
Micro and Nanoelectronics
Nanomaterials
Organic Electronics
Polymer Science
Key Resources
Stable Isotopes (ISOTEC)
 SOLID POLYMERIC ELECTROLYTES (SPE)
Energy Source Materials
Solid polymeric electrolytes (SPEs) are used as polymer electrolyte membranes in PEM fuel cells.
fuel cell schematic
Schematic of a typical proton exchange membrane (PEM) fuel cell.
Cross-linkable sulfonated SPE formulations  
Poly(styrene-ran-ethylene)659401  Poly(styrene-ran-ethylene), sulfonated, cross-linkable, 5 wt.% solution in 1-propanol
  • Contains <0.5% vinyl silane cross-linking agent
  • Styrene 76 wt.%
  • Sulfonated styrene units 32-38%
  • Conductivity = 0.1184 S/cm
  • The ionic conductivity can increase up to 50% when heating from RT to 67 °C.
  • Treat with 0.5 M Sulfuric acid for 20 min to strengthen after cross-linking.
  • Material becomes cross-linked w/800-1000 mJ/cm2 UV exposure after drying at 110°C for 10-15 min; or at 140-150°C for 1 min.
Polystyrene-block-poly(ethylene-ran-butylene)block-polystyrene659444  Polystyrene-block-poly(ethylene-ran-butylene)block-polystyrene, sulfonated, cross-linkable, 5 wt.% solution in 1-propanol
  • Contains <0.5% vinyl silane cross-linking agent
  • Styrene 29 wt.%
  • Sulfonated styrene units 55-65%
  • Conductivity = 0.1078 S/cm
  • The ionic conductivity can increase up to 50% when heating from RT to 67 °C.
  • Treat with 0.5 M Sulfuric acid for 20 min to strengthen after cross-linking.
  • Material becomes cross-linked w/800-1000 mJ/cm2 UV exposure after drying at 110°C for 10-15 min; or at 140-150°C for 1 min.
Other sulfonated SPE formulations  
Poly(styrene-ran-ethylene)659592  Poly(styrene-ran-ethylene), sulfonated, 5 wt.% solution in 1-propanol
  • Styrene 76 wt.%
  • Sulfonated styrene units 32-38%
  • Conductivity = 0.1494 S/cm
  • The ionic conductivity can increase up to 50% when heating from RT to 67 °C.
Polystyrene-block-poly(ethylene/butylene)-block-polystyrene448885  Polystyrene-block-poly(ethylene/butylene)-block-polystyrene, sulfonated solution, 5 wt.% solution in propanol and dichlorethane
  • Styrene 29 wt.%
  • Sulfonated styrene units 55-65%
  • Conductivity ~ 0.07 S/cm
A spin coat system is recommended for casting these polymers into films. Click here to view the tech bulletin on the spin coat system. (78.2 KB PDF)
Spin-Coating Systems  
Product Name Product # 
Chemat precision spin-coater, 2-stage, 220V Z551589
Chemat precision spin-coater, 2-stage, 115V Z551562
Manual Film-Casting Instructions
To cast a film without using a spin coat system, the following procedure is recommended:
1.  Concentrate solution to 600 centapoise viscosity (typically 6-9 wt. % polymer).
2.  Preheat the casting surface to 35°C. A clean, flat teflon-coated glass surface is recommended.
3.  Secure a silicone release liner to the casting surface.
4.  Place a small drop of the polymer solution on the release liner.
5.  Draw the polymer solution down the release liner.
6.  Dry the film for 30 min. For controlled evaporation it is recommended to cover surface with aluminum foil that has been perforated with small pinholes.