|
Packaged in steel cylinders designed for the Cambridge Nanotech Inc.
ALD system
| Product # |
Product Name |
Formula |
Physical State |
Add
to Cart |
|
679798 |
Bis(ethylcyclopentadienyl)ruthenium(II) |
Ru(C5H4C2H5)2 |
Liquid |
|
|
668885 |
Bis(tert-butylimino)bis(dimethylamino)tungsten(VI) |
((CH3)3CN)2W(N(CH3)2)2 |
Liquid, air-sensitive |
|
|
668729 |
Diethylzinc |
Zn(C2H5)2 |
Liquid, pyrophoric |
|
|
663301 |
Trimethylaluminum |
Al(CH3)3 |
Liquid, pyrophoric |
|
|
669008 |
Tetrakis(dimethylamido)titanium(IV) |
Ti[N(CH3)2]4 |
Liquid, air-sensitive |
|
|
687502 |
Titanium(IV) isopropoxide |
Ti(OPr-i)4 |
Liquid, air-sensitive |
|
|
669016 |
Tetrakis(dimethylamido)zirconium(IV) |
Zr[N(CH3)2]4 |
Solid, air-sensitive |
|
|
688509 |
Silicone tetrachloride |
SiCl4 |
Liquid, moisture-sensitive |
|
|
666610 |
Tetrakis(dimethylamido)hafnium(IV) |
Hf[N(CH3)2]4 |
Solid, air-sensitive |
|
|
668990 |
Tris(diethylamido)(tert-butylimido)tantalum |
(CH3)3CN=Ta(N(C2H5)2)3 |
Liquid, air-sensitive |