| Product # |
Product Name |
Synonym |
HLB |
Add to Cart |
|
278386 |
2,4,7,9-Tetramethyl-5-decyne-4,7-diol |
|
4.0 |
|
|
435406 |
PEG-block-PPG-block-PEG, MN 1100 |
|
4.0 |
|
|
435422 |
PEG-block-PPG-block-PEG, MN 2000 |
|
4.0 |
|
|
435430 |
PEG-block-PPG-block-PEG, MN 2800 |
|
4.0 |
|
|
435457 |
PEG-block-PPG-block-PEG, MN 4400 |
|
4.0 |
|
|
435511 |
Ethylenediamine tetrakis(PO-b-EO)
tetrol, MN 3600 |
|
4.0 |
|
|
435546 |
Ethylenediamine tetrakis(EO-b-PO)
tetrol, MN 7200 |
|
4.0 |
|
|
435554 |
Ethylenediamine tetrakis(EO-b-PO)
tetrol, MN 8000 |
|
4.0 |
|
|
238562 |
Igepal® CA-210 |
Polyoxyethylene(2)
isooctylphenyl ether |
4.3 |
|
|
388912 |
Sorbitan monooleate |
Span® 80 |
4.3 |
|
|
435503 |
PPG-block-PEG-block-PPG, MN 3300 |
|
4.5 |
|
|
238635 |
Igepal CO-210 |
Polyoxyethylene(2)
nonylphenyl ether |
4.6 |
|
|
318221 |
Sorbitan monostearate |
Span® 60 |
4.7 |
|
|
388866 |
Brij® 92 |
Polyoxyethylene(2)
oleyl ether |
4.9 |
|
|
388882 |
Brij® 72 |
Polyoxyethylene(2)
stearyl ether |
4.9 |
|
|
388831 |
Brij® 52 |
Polyoxyethylene(2)
cetyl ether |
5.3 |
|
|
388920 |
Sorbitan monopalmitate |
Span® 40 |
6.7 |
|
|
421286 |
Merpol® A surfactant |
|
6.7 |
|
|
461180 |
2,4,7,9-Tetramethyl-5-decyne-4,7-diol ethoxylate |
|
8.0 |
|
|
494755 |
Triton® SP-135 |
|
8.0 |
|
|
318213 |
Sorbitan monolaurate |
Span® 20 |
8.6 |
|
|
435465 |
PEG-block-PPG-block-PEG, MN 5800 |
|
9.5 |
|
|
435481 |
PPG-block-PEG-block-PPG, MN 2700 |
|
9.5 |
|
|
235989 |
Brij® 30 |
Polyoxyethylene(4)
lauryl ether |
9.7 |
|
|
238570 |
Igepal® CA-520 |
Polyoxyethylene(5)
isooctylphenyl ether |
10.0 |
|
|
238643 |
Igepal® CO-520 |
Polyoxyethylene(5)
nonylphenyl ether |
10.0 |
|
|
466409 |
Polyoxyethylene sorbitol hexaoleate |
|
10.2 |
|
|
421340 |
Merpol® SE surfactant |
|
10.5 |
|
|
388904 |
Tween® 85 |
Polyoxyethylene(20)
sorbitan trioleate |
11.0 |
|
|
452602 |
8-Methyl-1-nonanol
propoxylate-block-ethoxylate |
|
11.0 |
|
|
466395 |
Polyoxyethylene sorbitan tetraoleate |
|
11.4 |
|
|
369349 |
Triton® X-114 |
Polyoxyethylene(8)
isooctylphenyl ether |
12.4 |
|
|
388890 |
Brij® 76 |
Polyoxyethylene(10)
stearyl ether |
12.4 |
|
|
431281 |
Brij® 97 |
Polyoxyethylene(10)
oleyl ether |
12.4 |
|
|
421332 |
Merpol® OJ surfactant |
|
12.5 |
|
|
388858 |
Brij® 56 |
Polyoxyethylene(10)
cetyl ether |
12.9 |
|
|
421359 |
Merpol® SH surfactant |
|
12.9 |
|
|
521078 |
Tergitol® NP-9 |
Nonylphenol polyethylene glycol ether |
12.9 |
|
|
461199 |
2,4,7,9-Tetramethyl-5-decyne-4,7-diol
ethoxylate (5 EO/OH) |
|
13.0 |
|
|
494763 |
Triton® SP-190 |
|
13.0 |
|
|
542334 |
Igepal® CO-630 |
Polyoxyethylene(9)
nonylphenyl ether |
13.0 |
|
|
234729 |
Triton® X-100 |
Polyoxyethylene(10)
isooctylphenyl ether |
13.5 |
|
|
238651 |
Igepal® CO-720 |
Polyoxyethylene(12)
nonylphenyl ether |
14.2 |
|
|
466417 |
Polyoxyethylene(12) tridecyl ether |
|
14.5 |
|
|
466875 |
Polyoxyethylene(18) tridecyl ether |
|
14.5 |
|
|
238589 |
Igepal® CA-720 |
Polyoxyethylene(12)
isooctylphenyl ether |
14.6 |
|
|
274364 |
Tween® 80 |
Polyoxyethylene(20)
sorbitan monooleate |
14.9 |
|
|
374253 |
Tween® 60 |
Polyoxyethylene(20)
sorbitan monostearate |
15.0 |
|
|
435449 |
PEG-block-PPG-block-PEG, MN 2900 |
|
15.0 |
|
|
435473 |
PPG-block-PEG-block-PPG, MN 2000 |
|
15.0 |
|
|
236004 |
Brij® 78 |
Polyoxyethylene(20)
stearyl ether |
15.3 |
|
|
436240 |
Brij® 98 |
Polyoxyethylene(20)
oleyl ether |
15.3 |
|
|
421308 |
Merpol® HCS surfactant |
|
15.5 |
|
|
274356 |
Tween® 40 |
Polyoxyethylene(20)
sorbitan monopalmitate |
15.6 |
|
|
235997 |
Brij® 58 |
Polyoxyethylene(20)
cetyl ether |
15.7 |
|
|
525901 |
Polyethylene-block-poly(ethylene
glycol)Mn 2250 |
|
16.0 |
|
|
274348 |
Tween® 20 |
Polyoxyethylene(20)
sorbitan monolaurate |
16.7 |
|
|
858366 |
Brij® 35 |
Polyoxyethylene(23)
lauryl ether |
16.9 |
|
|
461202 |
2,4,7,9-Tetramethyl-5-decyne-4,7-diol ethoxylate (15 EO/OH) |
|
17.0 |
|
|
238678 |
Igepal® CO-890 |
Polyoxyethylene(40)
nonylphenyl ether |
17.8 |
|
|
234737 |
Triton® X-405 |
Polyoxyethylene(40)
isooctylphenyl ether |
17.9 |
|
|
466387 |
Brij® 700 |
Polyoxyethylene(100)
stearyl ether |
18.8 |
|
|
238686 |
Igepal® CO-990 |
Polyoxyethylene(100)
nonylphenyl ether |
19.0 |
|
|
238694 |
Igepal® DM-970 |
Polyoxyethylene(150)
dinonylphenyl ether |
19.0 |
|
|
435414 |
PEG-block-PPG-block-PEG, MN 1900 |
|
20.5 |
|
|
412325 |
PEG-block-PPG-block-PEG, MN 8400 |
|
24.0 |
|
|
435538 |
Ethylenediamine tetrakis(PO-b-EO)
tetrol, MN 15000 |
|
24.0 |
|
|
542342 |
PEG-block-PPG-block-PEG, average Mn
ca. 14,600 |
|
27.0 |