New Products
Nanotechnology
Biocompatible/Biodegradable
Display and Optoelectronics
Energy Source Materials
Metals and Ceramic Science
Micro and Nanoelectronics
Nanomaterials
Dendrimers
Lumidots
Nanoclays
Nanopowders
Nanotubes, Fullerenes
Tutorial
Polymer Science
Key Resources
Stable Isotopes (ISOTEC)
 Nano Minerals: Nanoclays
Nanomaterials
Nanoclay Products  |  Technical Notes  |  Related Products
Nanoclays are nanoparticles of layered mineral silicates. Depending on chemical composition and nanoparticle morphology, nanoclays are organized into several classes such as montmorillonite, bentonite, kaolinite, hectorite, and halloysite. Organically-modified nanoclays (organoclays) are an attractive class of hybrid organic-inorganic nanomaterials with potential uses in polymer nanocomposites, as rheological modifiers, gas absorbents and drug delivery carriers.
Montmorillonite Nanoclays
Plate-like montmorillonite is the most common nanoclay used in materials applications. Montmorillonite consists of ~ 1 nm thick aluminosilicate layers surface-substituted with metal cations and stacked in ~ 10 µm-sized multilayer stacks. Depending on surface modification of the clay layers, montmorillonite can be dispersed in a polymer matrix to form polymer-clay nanocomposite. Within the nanocomposite individual nm-thick clay layers become fully separated to form plate-like nanoparticles with very high (nm × µm) aspect ratio (Figure 1A). Sigma-Aldrich, in collaboration with the Nanocor Corporation, offers a range of montmorillonite nanoclay products with different organic modifications optimized to be compatible with various polymer systems.
Nanoclay Image   Figure 1:
A.
TEM micrograph of 2% nylon-clay nanocomposite showing nm-thick platelets of montmorillonite clay fully dispersed in the polymer matrix. Inset shows schematic of montmorillonite aluminosilicate layer structure.
B. TEM micrograph of halloysite clay nanotubes.
Halloysite Nanoclay
Halloysite is a naturally occurring aluminosilicate nanotube (Figure 1B). The two-layer halloysite tubes are chemically similar to kaoline and have average dimensions of 15 × 1000 nm comparable to carbon nanotubes. Halloysite tubes are hollow and can be used for controlled delivery and release of drugs as well as nanocomposite and rheology modification applications.
Technical Notes
Montmorillonite Nanoclays: product spotlight and selection table.
Polymer-Clay Nanocomposites: read technical tutorial by Dr. Alexander Morgan in Material Matters 2.1 (2.18 Mb PDF).
Halloysite Nanotubes: encapsulation and sustained release of macromolecules.
Related Products
Polymers
Nanoparticles
Biocompatible Materials