| Single Crystal Substrates |
|
Composition |
Formula |
Orientation |
Crystal Type |
Unit Cell (Å) |
Melting Point(°C) |
Density (g/cm3) |
Hardness (Moh) |
Product # |
|
Aluminum oxide single crystal substrate 99.99 % |
Al2O3 |
(0001) |
Hexagonal |
a =
4.758, c = 12.992 |
2040
|
3.98 |
9 |
634875 |
|
Lanthanum aluminum oxide single crystal substrate |
LaAlO3 |
<100> |
Rhombohedral < 435 °C
Cubic > 435 °C |
a = 5.357, c = 13.22
a = 3.821
|
2080
2080 |
6.52
6.52
|
5.9
5.9
|
634735 |
|
LSAT* 99.9+ % single crystal substrate |
(LaAlO3)0.3(Sr2AlTaO6)0.7 |
<100> |
Cubic |
a = 3.868 |
1840 |
6.74 |
6.5 |
635050 |
|
Magnesium aluminate single crystal substrate 99.99 % |
MgAl2O4 |
<100> |
Cubic |
a = 8.083 |
2130 |
3.64 |
8 |
635073 |
|
Magnesium aluminate single crystal substrate 99.99 % |
MgAl2O4 |
<110> |
Cubic |
a = 8.083 |
2130 |
3.64 |
8 |
634840 |
|
Magnesium aluminate single crystal substrate 99.99 % |
MgAl2O4 |
<111> |
Cubic |
a = 8.083 |
2130 |
3.64 |
8 |
634832 |
|
Magnesium oxide 99.9+ % single crystal substrate |
MgO |
<100> |
Cubic |
a = 4.216 |
2852 |
3.58 |
5.8 |
634646 |
|
Magnesium oxide single crystal substrate |
MgO |
<110> |
Cubic |
a = 4.216 |
2852 |
3.58 |
5.8 |
634700 |
|
Magnesium oxide single crystal substrate |
MgO |
<111> |
Cubic |
a = 4.216 |
2852 |
3.58 |
5.8 |
634697 |
|
Silicon dioxide single crystal substrate 99.99 % |
SiO2 |
(0001) |
Hexagonal |
a = 4.914, c = 5.405 |
1610 |
2.684 |
7 |
634867 |
|
Strontium lanthanum aluminate single crystal substrate 99.99 % |
SrLaAlO4 |
<001> |
Tetragonal |
a = 3.756, c = 12.63 |
1650 |
5.924 |
|
634891 |
|
Strontium lanthanum aluminate single crystal substrate 99.9+ % |
SrLaAlO4 |
<100> |
Tetragonal |
a = 3.756, c = 12.63 |
1650 |
5.924 |
|
635111 |
|
Strontium titanate single crystal substrate |
SrTiO3 |
<100> |
Cubic |
a = 3.905 |
2080 |
5.175 |
6 |
634689 |
|
Strontium titanate single crystal substrate |
SrTiO3 |
<110> |
Cubic |
a = 3.905 |
2080 |
5.175 |
6 |
634670 |
|
Strontium titanate 99.99 % single crystal substrate |
SrTiO3 |
<111> |
Cubic |
a = 3.905 |
2080 |
5.175 |
6 |
638161 |
|
Titanium(IV) oxide single crystal substrate 99.99 % |
TiO2 |
<001> |
Tetragonal |
a =4.5936, c = 2.9582 |
1840 |
4.26 |
7 |
635057 |
|
Titanium(IV) oxide single crystal substrate 99.99 % |
TiO2 |
<100> |
Tetragonal |
a =4.5936, c = 2.9582
|
1840 |
4.26 |
7 |
635049 |
|
Titanium(IV) oxide single crystal substrate 99.99 % |
TiO2 |
<110> |
Tetragonal |
a =4.5936, c = 2.9582 |
1840 |
4.26 |
7 |
635065 |
|
Yttrium vanadate single crystal substrate 99.99 % |
YVO4 |
<110> |
Tetragonal |
a = 7.12, c = 6.29 |
1825 |
4.22 |
5 |
635103 |
|
*LSAT=Lanthanum Strontium Aluminum Tantalum oxide |