New Products
Nanotechnology
Biocompatible/Biodegradable
Display and Optoelectronics
Energy Source Materials
Metals and Ceramic Science
Micro and Nanoelectronics
Nanomaterials
Polymer Science
New Products
Additives
Applications
Engineering Polymers
Free Radical Initiators
Functional Polymers
Initiators
Monomers
Polymer Analysis
Specialty Areas
Key Resources
Stable Isotopes (ISOTEC)
 FREE RADICAL INITIATORS
Polymerization Tools
  Photoinitiators
  Thermal Initiators
  Initiator/Stabilizer FAQs
Photoinitiators: Classification
A photoinitiator is a compound especially aded to a formulation to convert absorbed light energy, UV or visible light, into chemical energy in the form of initiating species, viz, free radicals or cations. Based on the mechanism by which intiating radicals are formed, photoinitiators are generally divided into two classes:
  • Type I photoinitiators undergo a unimolecular bond cleavage upon irradiation to yield free radicals.
  • Type II photoinitiators undergo a biomolecular reaction where the excited state of the photoinitiator interacts with a second molecule (a coinitiator) to generate free radicals.
UV photointiators of both Type I and Type II are available. However, visible light photoinitiators belong almost exclusively to the Type II class of photoinitiators. Table I summarizes the various classes of available Type I and Type II photoinitiators and their common applications.
Table I: Common Applications for Different Classes of Photoinitiators
UV-Photoinitiators
Visible Photoinitiators
Type I  Photoinitiators
Type II
Photoinitiator Class
Benzoin ethers
Benzil ketals
α-Dialkoxy-aceto-phenones
α-Hydroxy-alkyl-phenones
α-Amino- alkyl-phenones
Acyl-phosphine oxides
Benzo-phenones/ amines
Thio-xanthones/ amines
Titanocenes
Application
Wood coatings (polyester)
x
x
x
  x
Paper coatings
x
x
Clear coatings
(on metal, wood, plastic)
x
x
x
     x2
Printing Plates
x
x
    x
x
Offset inks
x1
x
x
x
Screen inks
    x3
x
x
Pigmented coatings
   x
x
White lacquers
    x4
  x
Photo resists
    x3
x
x
(1) In combination with benzophenone. (2) Only systems of high reactivty. (3) Partially in combination with thioxanthones. (4) Thin layers only.
This chart is reprinted with permission from SITA Technology Ltd.
Photoinitiators
Product #  Product Name Add to Cart
A10701 Acetophenone ReagentPlus, 99%
415952 Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide 97%
A88409 4,4′-Dimethoxybenzoin 95%
A90004 Anthraquinone 97%
123242 Anthraquinone-2-sulfonic acid Sodium salt 97%
119318 Benzene-chromium(0) tricarbonyl 98%
B5151 4-(Boc-aminomethyl)phenyl isothiocyanate ~95%
B5151 Benzil 98%
399396 Benzoin purified by sublimation, ≥99.5%
172006 Benzoin ethyl ether 99%
195782 Benzoin isobutyl ether technical grade, 90%
B8703 Benzoin methyl ether 96%
B9300 Benzophenone ReagentPlus, 99%
B9300 Benzoic acid meets USP testing specifications
405620 Benzophenone/1-hydroxycyclohexyl phenyl ketone, 50/50 blend
262463 Benzophenone-3,3′,4,4′-tetracarboxylic dianhydride 98%, purified by sublimation
B12601 4-Benzoylbiphenyl 99%
405647 2-Benzyl-2-(dimethylamino)-4′-morpholinobutyrophenone 97%
160326 4,4′-Bis(diethylamino)benzophenone ≥99%
147834 Michler’s ketone 98%
124893 (±)-Camphorquinone 97%
C72404 2-Chlorothioxanthen-9-one 98%
D31737 5-Dibenzosuberenone 97%
227102 2,2-Diethoxyacetophenone >95%
D110507 4,4′-Dihydroxybenzophenone 99%
196118 2,2-Dimethoxy-2-phenylacetophenone 99%
149349 4-(Dimethylamino)benzophenone 98%
146706 4,4′-Dimethylbenzil 97%
D149675 3,4-Dimethylbenzophenone 99%
405663 Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide/2-hydroxy-2-methylpropiophenone, blend
275719 4′-Ethoxyacetophenone 98%
E12206 2-Ethylanthraquinone ≥97%
F408 Ferrocene 98%
328103 3′-Hydroxyacetophenone ≥99%
278564 4′-Hydroxyacetophenone 99%
220434 3-Hydroxybenzophenone 99%
H20202 4-Hydroxybenzophenone 98%
405612 1-Hydroxycyclohexyl phenyl ketone 99%
405655 2-Hydroxy-2-methylpropiophenone 97%
157538 2-Methylbenzophenone 98%
198056 3-Methylbenzophenone 99%
M30507 Methyl benzoylformate 98%
405639 2-Methyl-4′-(methylthio)-2-morpholinopropiophenone 98%
156507 9,10-Phenanthrenequinone ≥99%
290742 4′-Phenoxyacetophenone 98%
T34002 Thioxanthen-9-one 97%
407216 Triarylsulfonium hexafluorophosphate salts, mixed 50% in propylene carbonate
405736 3-Mercapto-1-propanol 95%
447528 11-Mercapto-1-undecanol 97%
328375 1-Mercapto-2-propanol 95%
264792 3-Mercapto-2-butanol, mixture of isomers 97%
Thermal Initiatorsback to top
Product #  Product Name Solvent T(°C) kd(s-1) 10th Half-life °C(Solvent) Add to Cart
118168 4,4′-Azobis(4-cyanovaleric acid) ≥75% Acetone
Water
Water
70
69
80
4.6x10-5
1.9x10-5
9.0x10-5
69 (water)
380210 1,1′-Azobis(cyclohexanecarbonitrile) 98% Toluene 80
95
102
6.5x10-5
5.4x10-5
1.3x10-4
88 (toluene)
441090 2,2′-Azobis(2-methylpropionitrile) 98% Benzene 50
70
100
2.2x10-6
3.2x10-5
1.5x10-3
65 (toluene)
179981 Benzoyl peroxide reagent grade, 97% Benzene 60
78
100
2.0x10-6
2.3x10-5
5.0x10-4
70 (benzene)
441694 2,2-Bis(tert-butylperoxy)butane Solution 50 wt. % in mineral oil       100 (benzene)
388092 2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane technical grade, 90% Benzene 93 1.9x10-5 120 (benzene)
441716 Bis[1-(tert-butylperoxy)-1-methylethyl]benzene 96%   115
145
1.1x10-5
4.7x10-4
115 (benzene)
416665 tert-Butyl hydroperoxide Solution 5.0-6.0 M in decane Benzene 130
160
170
183
3x10-7
6.6x10-6
2.0x10-5
3.1x10-5
170 (benzene)
388076 tert-Butyl peracetate Solution 50 wt. % in odorless mineral spirits Benzene 85
100
130
1.2x10-6
1.5x10-5
5.7x10-4
100 (benzene)
168521 tert-Butyl peroxide 98% Benzene 80
100
130
7.8x10-8
8.8x10-7
3.0x10-5
125 (benzene)
159042 tert-Butyl peroxybenzoate 98% Benzene 100
130
1.1x10-5
3.5x10-4
103 (benzene)
247502 Cumene hydroperoxide technical grade, 80% Benzene 115
145
4.0x10-5
6.6x10-4
135 (toluene)
329541 Dicumyl peroxide 98% Benzene     115 (benzene)
290785 Lauroyl peroxide 97% Benzene 40
60
85
4.9x10-8
9.2x10-7
3.8x10-5
65 (benzene)
269336 Peracetic acid Solution 32 wt. % in dilute acetic acid       135 (toluene)
216224 Potassium persulfate ACS reagent, ≥99.0% Water
0.1M NaOH
80
50
60
80
90
6.9x10-8
9.5x10-7
3.2x10-6
9.2x10-5
3.5x10-4
60 (H2O)
70 (0.1M NaOH)
Back to top
Initiator/Stabilizer FAQ
 Dr. S.S. Newaz
Polyorganix Inc.
Houston, TX
Q. How does one choose the appropriate initiator?
               In a free radical addition polymerization, the choice of polymerization initiator depends mainly on two factors: a) its solubility and b) its decomposition temperature.  If the polymerization is performed in an organic solvent, then the initiator should be soluble in that solvent and the decomposition temperature of the initiator must be at or below the boiling point of the solvent. Commonly, 2,2′-Azobis(2-methylpropionitrile) (441090)  and 2-(4-Biphenyl)-5-phenyloxazole (216984) suit these requirements.  If the desired polymerization occurs at or below 20 °C, then special, low temperature free radical initiators need to be used.  Various AZO- type initiators can be chosen for range of decomposition temperature requirement.
               For emulsion polymerization or polymerization in an aqueous system, a water soluble initiator like potassium persulfate (379824) or an organic, water-soluble initiator 4,4′-Azobis(4-cyanovaleric acid) (118168) would be suitable.
Q. How does one determine the reactivity of monomer?
               Determination of monomer reactivity is not always obvious or straightforward.  Researchers rely on their experience and published data on individual monomers.  In general, extent of conjugation in the molecular structure can be viewed as indicative of its tendency to form the initial free radical required for propagating a free radical polymerization.  Usually, a more conjugated system is more likely to undergo the free radical polymerization.
Q. When is it necessary to remove a stabilizer prior to polymerization and how does one do so?
               Usually, it is not necessary to remove stabilizers.  They are typically present in ppm level and the use of a free radical initiator at the polymerization temperature will overwhelm the effect of the stabilizer.  In worst-case scenarios, one may need to add extra amounts of initiator to sustain an acceptable polymerization rate.  In most cases, once a polymerization initiates, the rate can be sustained without much difficulty.  If, however, it is absolutely necessary to remove the stabilizer, column chromatography is the preferred method.
Q. How does one remove residual initiator, stabilizer, and/or unreacted monomer after polymerization?
              It is a common practice to dissolve the polymers in a solvent prior to end use, followed by precipitating the polymer using a co-solvent.  Usually the residual initiators and stabilizers will remain in solution and the polymers will separate out as a solid (powder, gum or fibers).  This process may be repeated until desirable polymer characteristics are obtained.  This fractional precipitation is also effective in removing lower molecular weight polymers, resulting in narrower molecular weight distribution – of course accompanied with a loss of yield.  Typical solvent/co-solvent pairs could be: Toluene/hexane, toluene/methanol, THF/water, etc.