New Products
Nanotechnology
Biocompatible/Biodegradable
Energy Source Materials
Metals and Ceramic Science
Micro and Nanoelectronics
Nanomaterials
Organic Electronics
Polymer Science
Applications
Free Radical Initiators
Key Resources
Stable Isotopes (ISOTEC)
 ANALYSIS BY NMR
Materials Science
Polymer Analysis by NMR
Copolymer Analysis by NMR
Featured Polymer Products
Polymer Analysis by NMR
One of the challenges polymer scientists face is molecular weight (avg. chain length) determination of their materials. While membrane osmometry, gel permeation chromatography, viscosity analysis and mass spectrometry are typically used for molecular wt. determination, the techniques can be time consuming, inaccurate for the molecular wt. ranges involved or require specialized instrumentation. End group analysis by NMR offers an easy alternative method using an instrument commonly found in many analytical labs. In addition, NMR analysis can also be used to accurately determine monomer ratios for various copolymer molecules.
Scientists at Sigma-Aldrich routinely determine number-average molecular weight (Mn) by 1H NMR end-group analysis for polymers having Mn values under 3,000. Sensitivity of the instrument and the subsequent ability to detect end-group protons will determine the upper limit that can be measured. The method relies on a few simple needs.
  • Identifiable end-group protons distinguishable from repeating monomer group protons by NMR
  • Accurate integration of both end-group and monomer protons
  • Knowledge of monomer formula weights
Once the ratio of protons on the end-groups to protons on the polymer chain is determined, using the NMR, simple math can be applied to generate the Mn value.
The following example illustrates this method:
1) Calculation, integral per proton:
                       Locate the end group proton signals (ca. 5.8, 6.2 & 6.4 ppm)
integral per proton  =             sum of vinyl proton integrals            
                                    # of protons in the two vinyl end group
                                                            ß
 =           10.00 + 9.66 + 10.17
                                                                                    6
                                                                                    ß
     =               4.97 per proton
2) Calculation, number of repeating monomer units, n:
Locate the OCH2CH2 proton signals (ca. 3.6, 3.7 & 4.3 ppm)
n  =             (sum of methylene proton integrals)  /  # of methylene protons
                          {previously calculated integral per proton value}
                                                         ß
    =                                     (20.79 + 151.87) / 4  
                                                       4.97
                                                         ß
                            =                             8.69 repeating monomer units, n
3) Calculation, Mn:
Mn  =  (FW end groups) + (FW repeating unit)(n)
ß
                   =  (55.06 + 71.60) + (44.05)(8.69)
                                    ß                                
                   =            509
Copolymer Analysis by NMR
1H NMR is also a useful tool for calculating the monomer ratio in copolymers. As the following example illustrates, this method can be used even in situations where repeating unit proton signals overlap, as long as there is a clearly distinguishable signal from one of the repeating units.
1) Calculation, relative # of moles of propylene glycol (PG):
Locate the signal representing the methyl (CH3) protons (ca. 1.0 ppm)
Relative moles of polypropylene      =  integral for methyl protons
                                                          # of methyl group protons           
                                                                           ß
                                                      =               30.00
                                                                           3
                                                                           ß
                                                                              =  10.00 relative moles PG
2) Calculation, relative moles of ethylene glycol (EG):
Note:        There are 3 PG proton signals (representing the propylene glycol CH & CH2 groups) in the 3.0-4.0 ppm region overlapping the ethylene glycol methylene (CH2) protons in the same region.  The integral for these 3 PG protons will be equal to the integral representing the 3 previously identified propylene glycol methyl (CH3) protons.  In order to determine the molar ratio from the EG units, the PG integral must be subtracted from the total integral in the 3.0-4.0 ppm region.  
Locate the signals for the methylene protons (between 3.2 & 3.7 ppm)
            Relative moles of EG  =  sum of CH2 protons integral – CH3 protons integral
             # of EG protons
                                                                       ß
                                  =             (31.50 + 18.76 + 13.10) – 30.00  
                                                                       4
                                                                       ß
                                  =                    8.34 relative moles EG
3) Calculation, mole % ethylene glycol:
 mole % EG  =                relative moles EG     x   100%            
                              relative moles EG + relative moles PG      
                                                         ß
                    =                           8.34       x 100% 
                                          8.34 + 10.00
                                                         ß
                    =                45.5 mole % ethylene glycol
   Product Information
Product Name Product # 
Poly(ethylene glycol) diacrylate 437441
Poly(ethyleneglycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) average Mn ~5,800) 435465