|
All products are minimum 99% CP unless otherwise indicated.
Now you can order all of your PET products from one source.
Oxygen-18 is one of the key isotopes because it is the precursor of fluorine-18 used in PET. (Positron Emission Tomography). The most widely used of these, 18 F-Fluoro-D-glucose (18F-FDG), measures glucose metabolism and is widely applied in the detection and staging of malignancies, heart disease, epilepsy and other syndromes in clinical patients. PET scanning with 18F-labelled compounds, including FDG, is also used in many medical and drug research protocols. |
We offer an abundant supply of this important isotope in a variety of chemical forms with enrichments up to 99 atom %. We supply a wide variety of cyclotron target materials including Water-18O and Nitrogen-15N2/Oxygen gas mix, and we maintain an inventory of the most frequently requested products for rapid delivery. |
Please call us to discuss your needs. We can offer you significant savings in package deals.
Positron Emission Tomography Your Ultimate Source for Materials for the Production of 18FDG