Materials Science
Stable Isotopes (ISOTEC)
Biomolecular NMR
Biomolecular MS
MRI/MRS
Nutrition/Metabolism
PET
Agriculture
Custom Synthesis
NMR Solvents
Product Lines
New Products
Request A Quote
Key Resources
Customer Feedback
Customer Service
Gas/Gas Mixture
 NMR SOLVENTS
High Quality NMR Solvents
All products are minimum 99% CP unless otherwise indicated.
Aldrich provides a wide range of solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available. The highest deuterium enrichment solvents are available exclusively from Aldrich. We are alone in offering 13C-depleted solvents, useful when identifying signals of interest located in the same region of the spectrum as the 13C solvent's peaks. Aldrich now offers a complete line of NMR Reference Standards, which are fully tested to assure that the samples meet all NMR performance requirements. In addition to solvents Aldrich/Isotec offers a wide variety of compounds for NMR studies. These include 13C labeled glucose and 15N enriched ammonium salts for protein synthesis, a wide variety of compounds labeled with NMR active nuclei, and custom synthesis of unique compounds.
Test our High Quality NMR Solvents!   Ask your sales representative for a sample today.
Product Lines
    NMR Solvents and Reagents
Other Products of Interest
Shigemi NMR tubes, cleaners and accessories
NMR Books and Software
Spectral Viewer
Key Resources
Literature & Catalog Request
Equipment, Supplies & Books
Email to friend
Customer Feedback
Product Search
Browse Products
Frequently Asked Questions
Informational Flyer
"Double Twin Peaks" in H NMR of Deuterated Solvents QC/Analytical Isotec, Sigma-Aldrich (90 kb)
NMR Reference Standards       (22 Kb PDF)
Contact Us