| |
MRI/MRS |
 |
Magnetic Resonance Imaging (MRI) and Magnetic Resonance Spectroscopy (MRS) |
 |
 |
|
All products are minimum 99% CP unless otherwise indicated.
Use of 13C Isotopes in MR Research
Dr. Pratip Bhattacharya California Institute of Technology and Huntington Medical Research Institutes, Pasadena, California |
| Magnetic Resonance Imaging (MRI) and Magnetic Resonance Spectroscopy (MRS) are both valuable and highly accurate means for measuring and imaging metabolic processes in vivo using amino acids, carbohydrates, fatty acids or nucleic acids labeled with stable isotope like Carbon-13 (13C), Nitrogen-15 (15N), Phosphorus-31 (31P), Sodium-23 (23Na), Deuterium (H2), and Oxygen-17 (17O). These non-radioactive, stable isotopically labeled products allow for real-time imaging and quantification of metabolic process in vivo in plant, animal, or human subjects. |
13C MRS : The power of 13C MRS lies in its unique chemical specificity, enabling detection and quantification of metabolic intermediates which would not be so readily monitored using conventional radiochemical techniques. Improvements in NMR technologies, now permits us to obtain in vivo localized (1) and non-localized (2) 13C NMR spectra from rodent and human brain with similar quantity to those obtained earlier only under in vitro conditions, providing in this way a wealth of information on neurotransmitter recycling, cerebral bioenergetics in situ. In vivo and in vitro13C MRS was also accomplished in liver, skeletal muscle, heart, adipose tissue, kidney and pancreatic islets yielding a variety of invaluable information. Go to Article |
Product Lines
MRI/MRS