Order Center
Equipment, Supplies & Books
Corning Products
Bottle Top Vacuum Filters
Corning Glassware
Corning Plasticware
HYPERFlask
Product Lines
Products by Application
Product Highlights
Books
Gases Equipment
Glassware Catalog
Key Resources
Labware Literature
New Products
 Corning® HYPERFlask™ Cell Culture Vessel
Labware
For over 90 years, Corning® Life Science has been the industry leader in the manufacturing of high-quality cell culture supplies. With the release of the new Corning HYPERFlask cell culture vessel, Corning has brought new innovation to Cell Culture Plastics. Designed for high-yield applications, the Corning HYPERFlask reduces incubator space by offering the same combined growth area as 10 traditional 175 cm² flasks. The Corning HYPERFlask cell culture vessel has the same external dimensions as a standard T-175 cm² flask with 10 times the cell yield!
Learn how to use the Corning HYPERFlask. (398 Kb PDF)
HYPERFlask Cell Culture Vessel
Optimal Growth—Corning® CellBIND® surface treated gas permeable polystyrene for superior cell attachment and growth
Increase Cell Yield—10-fold higher cell yields increase productivity and capacity and reduces processing time and incubator space
Innovative Design—10 interconnected polystyrene growth surfaces
Save Time and Space—Reduce processing time and incubator storage space by handling one flask as compared to 10 traditional 175 cm² flasks
Automation Compatible—Bar coded and compatible with The Automation Partnership SelecT™ cell culture systems with an upgrade
Corning HYPERFlask Cell Culture Vessel Side View
A side view of the Corning HYPERFlask
shows the 10 gas permeable layers
that allow air exchange between
each layer.
Corning HYPERFlask Cell Culture Vessel and Traditional Flask
The Corning HYPERFlask
Cell Culture Vessel occupies the same
incubator space as a traditional
T-175 cm² flask with a 10 time
greater yield in cells.
Click to view the Corning HYPERFlask product detail and ordering information!
Technical Literature