|
GASES & EQUIPMENT |
 |
Equipment, Books & Supplies |
 |
 |
| |
|
Matheson Laboratory Gas Generators |
|
Achieve greater operating convenience and long-term reliability with Hydrogen, Nitrogen and Zero Air Generators. For more information about these products select the appropriate link below. |
|
| |
Request additional technical information, pricing and ordering information on
Matheson Generators.
| |
Sigma-Aldrich Lecture-Bottle Stations Safe and Simple Systems for Dispensing Lecture-Bottle Gases | |
 |
|
Dispensing
lecture-bottle gases has never been safer or simpler using these portable
lecture-bottle stations. The premium quality components housed
within a station are matched to the application, fully assembled,
leak-tested, and ready for use. Just connect the station to the
lecture-bottle, process with a source of inert gas if using a station with
a purge valve and you are ready to go.
 |
Non-Corrosive Lecture-Bottle Station
For
Inert and non-corrosive gas applications
Z562599-1EA
High-Purity Lecture-Bottle Station
For non-corrosive, high-purity gas applications Z562602-1EA
|
| Lecture-Bottle Station with T-Purge Valve For mildly-corrosive and high-purity gas applications
Z562610-1EA
Purge valve sits between the lecture-bottle and regulator to purge
system with inert gas to remove air, moisture, and process gas to maintain
high-purity conditions and to minimize gas hydrolysis and corrosion damage
to equipment. Purge system with dry, inert gas before introducing
mildly corrosive gases and again immediately after use. |
 |
 |
Corrosive Lecture-Bottle Station with
Cross-Purge Valve
For strong, acid-forming gas applications
Z562629-1EA
For use with hydrogen chloride, hydrogen, bromide, hydrogen fluoride,
and silicon tetrafluoride. Cross-purge valve sits between the
lecture-bottle and regulator to prevent escape of toxic and flammable
gases when changing cylinders and permits purging system with inert gas to
remove air, moisture, and process gas to minimize gas hydrolysis and
corrosion damage to equipment. Purge system with dry, inert gas
before introducing corrosive gases and again immediately after use. |
|
Lecture-Bottle Service Station
Z562637-1EA
This station
provides storage capacity for three additional lecture-bottles, tools, and
supplies for use with Sigma-Aldrich Lecture-Bottle Stations. Table
top or wall-mountable. Hinged cover over storage space keeps
supplies clean and secure. |
 |
| |
|
 |
CGA adapters
Allows for adapting the fittings on the lecture-bottle stations to all other cylinder fittings.
|
CGA |
NPTM (in.) |
Prod No. |
|
350 |
¼ |
Z564494 |
|
660 |
¼ |
Z564508 |
|
330 |
¼ |
Z564486 |
|
170 |
¼ |
Z564478 |
| |
|
4-way Wrench
Z564516
One wrench to fit both CGA and NPTM fittings. |