Sigma-Aldrich
Labware logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Glassware Catalog > Specialty Adapters
Glassware Catalog
Specialty Adapters
Connecting  •  Reducing/Enlarging  •  Septum-Inlet  •  Specialty  •  Tubing  •  Vacuum
On this page: Luer lock adaptersPour-out AdaptersHigh-dilution adapter
Luer lock adapters
al_Z17356L
Permits fitting of standard glassware with needles for air-sensitive transfer.
Description
Catalog Number
S.T. 24/40F to male Luer lock
S.T. 24/40M to male Luer lock
   0-3mm PTFE needle valve and 1 1/2in (38mm) drip-tip.
Back to Top
Pour-out Adapters
al_glassware_note5
al_Z17295L
S.T. Joint
Catalog Number
14/20
19/22
24/40
29/32
Back to Top
High-dilution adapter
al_glassware_note6
al_Z22330L
S.T. 24/40 joints. Reactants added dropwise from an addition funnel are prediluted with approximately 25mL of refluxing solvent held in a reservoir by an internal collar. Described in Organic Synthesis, 1990, 69, 180.
Z22,330-1
Back to Top