|
Glassware Catalog
Tubing Adapters
|
On this page: Torion connection • W/ Stopcock • Accessories
Tubing adapters with Torion connection
|
|
- Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE- coated sealing ring of the adapter.
- Turn the bushing to make an airtight compression seal with the tubing.
- Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use.
|
Tubing o.d. (in.)
|
Inner S.T. Joint
|
Straight Catalog Number
|
With hose barb Catalog Number
|
|
3/16
|
14/20
|
3/16
|
19/22
|
5/16
|
14/20
|
5/16
|
19/22
|
5/16
|
24/40
|
5/16
|
29/32
|
1/2
|
24/40
|
1/2
|
29/32
|
1/2
|
34/45
|
1/2
|
45/50
 |
Back to Top
Tubing adapters with stopcock
| Glass and PTFE stopcocks are threaded for additional safety and have a 2mm bore for 3/16 in. tubing connection and 4mm bore for 5/16 in. and 1/2 in. tubing connections. |
 |
|
Tubing o.d. (in.)
|
Inner S.T. Joint
|
PTFE stopcock Catalog Number
|
Glass stopcock Catalog Number
|
|
3/16
|
14/20
|
3/16
|
19/22
|
5/16
|
14/20
|
5/16
|
19/22
|
5/16
|
24/40
|
5/16
|
29/32
|
1/2
|
24/40
|
1/2
|
29/32
|
1/2
|
34/45
|
1/2
|
45/50
|
Back to Top
ACCESSORIES
Solid glass plugs
| Replace transfer tubing with this plug to make a leakproof Torion seal. |
 |
|
Plug o.d. (in.)
|
Catalog Number
|
|
3/16
|
|
|
5/16
|
|
|
1/2
|
|
|
Replacement PTFE-coated sealed rings
|
I.d. (in.)
|
Catalog Number
|
|
3/16
|
|
|
5/16
|
|
|
1/2