|
| |
|
On this page:
Mini gas bubbler Reaction vessel
bubbler In-line bubbler
Reverse-flow bubbler In-line oil bubblers
Check-valve bubblers
Safety bubbler Mercury bubbler
Accessories
Mini gas bubbler
Back to Top
Reaction vessel bubbler
 |
Capacity 5 to 7mL.
|
S.
T. Joint
|
Catalog
Number
|
|
14/20
|
|
|
19/22
|
|
|
24/40
|
|
|
29/32
|
|
|
Back to Top
In-line bubbler
Back to Top
Reverse-flow bubbler
 |
Monitors gas flow
into (or gas evolution from) a system.
Capacity 25mL per chamber.
Z14,795-8 |
Back to Top
In-line oil bubblers
Connect reaction vessel to in-line S.T. joint
or use with a ballast bulb to keep pressure constant. Capacity 10mL.
 |
 |
|
S.T.
Joint
|
Catalog
Number
|
|
14/20
|
|
|
19/22
|
|
|
19/38
|
|
|
24/40
|
|
|
29/32
|
|
|
Back to Top
Check-valve bubblers
 |
Check-valve ball prevents oil from being drawn
into system.
Capacity 15mL.
|
Description
|
Catalog
Number
|
| Single-inlet
tube, top outlet |
|
| T-inlet
tube, side outlet |
|
|
Back to Top
Safety bubbler
 |
Built-in flash arrester bulb prevents backflow.
Capacity 15mL fill mark.
Z22,372-7 |
Back to Top
Mercury bubbler
 |
Permits increased pressure within reaction vessels.
Internal reservoir prevents mercury from being drawn into reaction
flask or pushed our of bubbler. Do not overfill with mercury.
S.T. 24/40 joint.
Capacity 40mL.
Z15,243-9
Mineral Oil
6 x12mL
16,140-3
|
Back to Top
Accessories
Tygon (Formula 2075) chemically resistant tubing clear, flexible tubing
suitable for specialty chemical production and hazardous material handling. Autoclavable.
- Resistant to MEK, acids, bases, ketones, and alcohols
- Plasticizer free no contamination from leaching
- Biocompatible meets USP Class VI criteria
- No disposal concerns releases only carbon dioxide and water
upon incineration
 |
|
I.d.
(in.)
|
Wall
(in.)
|
O.d.
(in.)
|
Catalog
Number
|
|
1/16
|
1/16
|
3/16
|
|
|
1/8
|
1/16
|
1/4
|
|
|
3/16
|
1/16
|
5/16
|
|
|
1/4
|
1/16
|
3/8
|
|
|
5/16
|
1/16
|
7/16
|
|
|
3/8
|
1/16
|
1/2
|
|
|
1/2
|
1/16
|
3/4
|
|
|
5/8
|
1/8
|
7/8