ALDRICH HPLC SOLVENT RESERVOIR SYSTEMS
On this page:
Four Valve Caps Three
Valve Caps
Three Hole Caps Accessories
Features:
- Safety coated with autoclavable PVC
- Protects light sensitive materials to 385nm
- Fingertip controls for easy, precise control
- Specially designed valves and connections assure leak free operation
- Low profile minimizes chance of tipping
- Reservoir indents improve mixing
- Reference graduations allow run time calculations
HPLC systems with four valve caps
Valves controlling
helium sparge, venting, vacuum and solvent delivery are conveniently located
on the front of the cap. All tubing connects to the back of the cap, minimizing
clutter. Uses standard 47mm diameter filter membranes. Available as standard
(PTFE/PP O-rings) or THF-resistant systems (Chemraz O-rings).
Each system includes:
- (1) High-performance HPLC reservoir
- (1) 4-valve cap
- (1) Solvent pickup adapter with PTFE tubing
- (10) 1/4-28 nuts for 1/8in. o.d. tubing
- (10) Ferrules for 1/8in. o.d. tubing
- (2) Vacuum adapters, 1/4-28 to 1/4in. i.d. tubing
- 10ft PTFE tubing, 1/8in. o.d.
- (2) 25m SS filters
- (1) PTFE coated stir bar
- (2) Adapters for 1/4-28 to 1/8in. i.d. tubing (for Waters HPLC
systems with 1/8in. tubing)
 |
 |
|
Cap.
(mL)
|
Dimensions
diam. x H (mm)
|
Standard
systems
Catalog Number
|
THF-resistant systems
Catalog Number
|
|
250
|
175
x 285
|
500
|
190
x 300
|
1,000
|
120 x 400
|
2,000
|
150 x 450
|
5,000
|
195 x 500
|
10,000
|
240 x 595
|
20,000
|
305 x 675
|
Back to Top
Economy HPLC systems with
three valve caps
These systems allow for the sharing of one filtration/degassing
cap assembly (sold separately on next page) with several solvent
reservoirs. A simple cap change is required between the filtration
and sparge/delivery operations. Available as standard (PTFE/PP
O-rings) or THF-resistant systems (Chemraz O-rings).
Each system includes:
- (1) High-performance HPLC reservoir
- (1) 3-valve delivery cap
- (8) 1/4-28 nuts for 1/8in. tubing
- (8) Ferrules for 1/8in. o.d. tubing
- 10ft PTFE tubing 1/8in. o.d.
- (2) 25m SS filters
- (1) PTFE coated stir bar
- (2) Adapters for 1/4-28 to 1/8in. i.d. tubing
(for Waters HPLC systems with 1/8in. tubing)
 |
 |
|
Cap.
(mL)
|
Dimensions
diam. x H (mm)
|
Standard systems
Catalog Number
|
THF-resistant
systems
Catalog Number
|
|
250
|
175
x 285
|
500
|
190
x 300
|
1,000
|
120 x 400
|
2,000
|
150 x 450
|
5,000
|
195 x 500
|
10,000
|
240 x 595
|
20,000
|
305 x 675
|
Back to Top
Economy HPLC systems with
three hole caps
This economy system is designed to the same exacting
performance standards as the economy system on the previous page.
Not recommended for THF or chlorinated hydrocarbons.
Each system includes:
- (1) High-performance HPLC reservoir
- (1) 3-hole delivery cap
- (4) 1/4-28 nuts for 1/8in. o.d. tubing
- (4) Ferrules for 1/8in. o.d. tubing
- (3) 1/4-28 solid plugs
- 10ft PTFE tubing, 1/8in. o.d.
- (2) 25m SS filters
- (1) PTFE coated stir bar
- (2) Adapters for 1/4-28 to 1/8in. i.d. tubing
(for Waters HPLC systems with 1/8in. tubing)
|
 |
|
Cap.(mL)
|
Dimensions
diam. x H (mm)
|
Catalog
Number
|
|
250
|
75 x 285
|
|
|
500
|
90 x 300
|
|
|
1,000
|
120 x 400
|
|
|
2,000
|
150 x 450
|
|
|
5,000
|
195 x 500
|
|
|
10,000
|
240 x 595
|
|
|
20,000
|
305 x 675
|
|
|
Back to Top
Accessories
Filtration/degassing cap
 |
Glass-filled PTFE construction with a quick turn
cap to secure funnel or solvent pick up adapter. Supplied complete
with 47mm fritted glass membrane support. For use with solvent pickup
adapter (see below).
Z52,899-4
|
Solvent pickup adapter with valve
 |
This all glass adapter is designed specifically
for use with the filtration/ degassing cap (above) and all Aldrich
HPLC solvent reservoir systems. This adapter holds a membrane
filter securely in place while providing for continuous in-line
filtration of mobile phase. Supplied complete with PTFE connector
and 3ft of 1/4in. PTFE tubing and 4mm PTFE stopcock.
Z52,900-1
|
HPLC membrane filters
- Nylaflo - nylon, for general-purpose applications, offers
good chemical resistance with low extractables
- PTFE - PTFE on PP support for better stability and easier
handling, chemically inert for aggressive organics, hydrophobic. Designed
to perform at high temperatures (up to 145°C)
|
Membrane
|
Diam
(mm)
|
0.2 mm
Catalog Number
|
0.45 mm
Catalog Number
|
1.0 mm
Catalog Number
|
|
Nylaflo
|
47
|
|
|
-
|
|
PTFE
|
47
|
For a full listing of filtration supplies please see the Aldrich Handbook.
Back to Top |