Order Center
Equipment, Supplies & Books
Corning Products
Product Lines
Products by Application
Product Highlights
Books
Gases Equipment
Glassware Catalog
Key Resources
Labware Literature
New Products
On this page:
Super condensersTriple-jacket coil condenserTriple-coil condensers
Airflux condenser apparatusCoiled reflux condensers
Aldrich jacketed Friedrichs coil condensersAldrich modified Friedrichs condensers
Dry ice condensersRotary evaporator condenserCold-finger condensers
Graham condensersHartzler condensersShort condenser
Liebig condensersModified Davies condensers
Super condensers
Excellent for volatile compounds.
Overall H
(cm)
Surface area (cm2)
O.d.
(mm)
S.T. 24/40 Joints
Catalog Number
S.T. 29/32 Joints
Catalog Number
32
405
42
37
510
42
41
610
42
51
820
42
Back to Top
Triple-jacket coil condenser
Triple jacketing provides maximum cooling surface area. Minimizes loss of volatile components.
Overall
H (cm)
Surface area
(cm2)
O.d.
(mm)
S.T. 24/40 Joints
Catalog Number
S.T. 29/32 Joints
Catalog Number
44
1,230
60
Back to Top
Triple-coil condensers
Large surface area provides efficient condensation of refluxing liquids with reduced risk of flooding. For use with flasks up to 72L.
Overall H
(cm)
Surface area (cm2)
O.d.
(mm)
Ball
Socket Joints
Catalog
Number
Single Stack, 43cm coil
61
3,650
75
65/40
61
3,650
75
75/50
Double Stack, two 43cm coil sets
107
7,015
75
65/40
107
7,015
75
75/50
Back to Top
Airflux condenser apparatus
Efficient aluminum heat sink disperses heat. Remove plastic cap on heat sink to fill with distilled water. Water passes through to glass condenser. Complete apparatus includes Airflux aluminum heat sink, removable glass condenser, and boiling flask.
  • Use with flasks up to 250mL size and heating mantles with 60W max. output
  • Condensers can be mounted in tandem* for flasks sizes up to 1L and 300W heating mantles with a controller for steady reflux rate
  • Mounts to 1/2in. lattice-rods or support stands via built-in clamp on heat sink
Airflux
size
Flaskcap.
(mL)
S.T. 24/40 joints
Catalog Number
S.T. 24/29 joints
Catalog Number
S.T. 29/32 joints
Catalog Number
Small
100
Large
250
*Note: Do not use tandem mounted water-cooled Airflux units to condense solvents boiling over 100°C. The lower unit works at a temperature close to the boiling point of the solvent at high reflux rates. Alternative coolants such as ethylene glycol may be suitable. Tests should be carried out to determine satisfactory performance.
Back to Top
Coiled reflux condensers
The coiled reflux condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Overall H (cm)
S.T. Joints
Standard
Catalog Number
Jacketed
Catalog Number
   10 *
14/20
20
14/20
20
24/40
20
29/32
30
14/20
30
24/40
30
29/32
40
24/40
40
29/32
50
24/40
50
29/32
60
24/40
60
29/32
* Jacket length.
Back to Top
Aldrich jacketed Friedrichs coil condensers
The molded spiral condensing tube around the inner coil creates additional surface area for improved efficiency. 54mm O.D.
Overall H (cm)
S.T. 24/40
Catalog Number
S.T. 29/32
Catalog Number
26
37
Back to Top
Aldrich modified Friedrichs condensers
Molded spiral condensing coolant tube fits closely within the jacket to force vapors along the spiral path.
Features:
  • Modified feed tube to reduce chance of breakage
  • Additional sizes to meet specific applications
Jacket O.D. (mm)
Overall H (mm)
S.T. Joint
Catalog Number
25
230
14/20
37
160
14/20
37
270
24/40
37
270
29/32
37
370
24/40
37
370
29/32
54
300
24/40
54
300
29/32
Back to Top
Dry ice condensers
For Büchi rotary evaporators R110, R114, R124 and R151.
For use with short-stem duct. 35/25 ball joint.
Description
Catalog Number
Trap with hose barb
Trap with high vacuum, hollow glass stopcock
35/20 ball joint. Modified feed tube allows recharging during stripping.
Z15,244-7
With S.T. 24/40 ground-glass joints.
Z10,309-8
Back to Top
Rotary evaporator diagonal condenser
35/25 side ball joint at 105° angle, and No. 50 flange. 37cm overall length.
Z22,477-4
Back to Top
Cold-finger condensers
31cm overall length. 25 L x 6cm diam. cold-finger.
S.T. Joint
Catalog Number
14/20
24/40
29/32
Back to Top
Graham condensers
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil.
Overall H (cm)
S.T. Joint
Catalog Number
 10*
14/20
20
14/20
20
24/40
20
29/32
30
14/20
30
24/40
30
29/32
40
24/40
40
29/32
50
24/40
50
29/32
60
24/40
60
29/32
* Jacket length.
Back to Top
Hartzler condensers
34cm column length. Lower primary condenser of bulbous portions connected with each other and with an upper curvilinear column. Designed by Harold E. Hartzler; Goshen, IN (U.S. Patent 4 333 523).
S.T. Joint
Catalog Number
19/38
24/40
29/42
Back to Top
Short condenser
With S.T. 24/40 ground-glass joints. Useful in distilling solids. May be used for conventional distillations as a condenser or a jacketed column. 17.5cm jacket length.
Z11,575-4
Back to Top
Liebig condensers
Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Overall H (cm)
S.T. Joints
Catalog Number
  6*
14/20
 10*
14/20
20
14/20
20
24/40
20
29/32
30
14/20
30
24/40
30
29/32
40
24/40
40
29/32
50
24/40
50
29/32
60
24/40
60
29/32
* Jacket length.
Back to Top
Modified Davies condensers
Additional internal cooling surface increases efficiency with negligible hold-up when used at a standard 75° angle.
Jacketed L (cm)
S.T. 24/40 joint
Catalog Number
S.T. 29/32 joint
Catalog Number
25
40