Sigma-Aldrich
Labware logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Glassware Catalog > Distillation Columns
Glassware Catalog
Distillation Columns
Apparatus  •  Columns  •  Packing  •  Heads  •  Traps
On this page:
Vigreux: MicroStandardSilvered, vacuum-jacketed
Miscellaneous: Distilling columnVacuum-jacketedSilvered bellowsSilvered coil
Hemple: Hemple columnsStrip-silveredOldershaw
Snyder: Aldrich Snyder distilling columns
Micro Vigreux columns
al_Z17578L With S.T. 14/20 joints.
Effective L (mm)
Catalog Number
Vacuum-jacketed, 6mm i.d., 15mm o.d.
75
150
300
Unjacketed, 10mm i.d., 12mm o.d.
75
150
190
300
* Fits Aldrich All-in-One Lab Glassware Kit
Back to top
Standard Vigreux columns
With S.T. 24/40 joints
Effective L (mm)
Catalog Number
Vacuum-jacketed, 19mm i.d., 38mm o.d.
50
75
150
300
Unjacketed, 19mm i.d., 22mm o.d.
50
75
150
300
Back to top
Silvered, vacuum-jacketed Vigreux columns
Vacuum-jacketed and strip silvered to provide two windows 180° apart for viewing. S.T. 24/40 joints, 19mm i.d., 38mm o.d.
L (mm)
Catalog Number
250
400
Back to top
MISCELLANEOUS COLUMNS
al_glassware_distill2
A. Distilling column
S.T. 19/22 Clear-Seal joints, 200mm L, 30mm o.d.
Z10,031-5
B. Vacuum-jacketed distilling column
al_glassware_note52
S.T. 24/40 joints, 300mm L, 38mm o.d.
Z10,259-8
C. Silvered bellows column
S.T. 24/40 joints, 280mm L, 64mm o.d. Vacuum-jacketed. Bellows handle expansion from heat generated in inner straight tube. Strip-silvered to provide two windows 180° apart for viewing.
Z17,351-7
D. Silvered coil column
S.T. 24/40 joints, 280mm L, 57mm o.d. Vacuum-jacketed. Strip-silvered to provide two windows 180° apart for viewing.
Z17,352-5
Back to top
ALDRICH HEMPEL-TYPE DISTILLING COLUMNS
Hemple columns
al_Z17573L
Effective L (mm)
Micro
S.T. 14/20 joints
Catalog Number
Standard
S.T. 24/40 joints
Catalog Number
Vacuum-jacketed
10mm i.d., 25 mm o.d.
19mm i.d., 38mm o.d.
75
150
300
Unjacketed
10mm i.d., 12 mm o.d.
19mm i.d., 22 mm o.d.
75
150
300
Back to top
Strip-silvered Hempel columns
Vacuum-jacketed and strip-silvered to provide two windows 180° apart for viewing (except the 160mm column which is completely silvered).
L (mm)
I.d. (mm)
O.d. (mm)
S.T. Joints
Catalog Number
160
12.5
25
14/20
135
19
42
24/40
250
19
42
24/40
385
19
42
24/40
500
19
42
24/40
Back to top
Oldershaw distilling columns
al_Z17567L
Vacuum jacketed. Strip-silvered to provide two windows 180° apart for viewing. Consists of a series of sealed-in, perforated plates, each of which has a drain tube to direct condensed distillate to next lower plate and a weir to maintain uniform liquid level.
No. of plates
I.d. (mm)
O.d. (mm)
S.T. Joints
Catalog Number
5
19
64
24/40
5
19
64
29/42
5
28
64
29/42
10
19
64
24/40
10
19
64
29/42
10
28
64
29/42
Back to top
ALDRICH SNYDER DISTILLING COLUMNS
al_Z51696L Floating ball type with S.T. joints, 12mm o.d.
S.T. Joints
Two Sections
Catalog Number
Three Sections
Catalog Number
Four Sections
Catalog Number
Five Sections
Catalog Number
Effective L (mm)
120
145
170
195
14/20
-
19/22
-
24/40
29/32
Back to top