|
Glassware Catalog
Distillation Heads
|
On this page: Concentrator condenser • Water cooled distilling heads Multi-purpose distillation heads • Anti-creep distilling heads Distilling adapters • Short-path distillation apparatus Short-path distillation head • Aldrich Claisen distillation adapter Claisen distillation heads • Fractional distillation head Aldrich heavy-duty Vigreux, jacketed distillation head Micro distillation head apparatus • Fraction cutter • Micro distillation head Quick-distillation and sample collection tube • Variable take-off splitter heads High-capacity variable take-off splitter head • Aldrich falling-film distillation heads Jacketed falling-film distillation head
Concentrator condenser
Vapors rise up the inner tube and condense on the highly efficient spiral path. Replaces traditional 3-piece distillation set-up.
|
S.T. Joints
|
Overall H (mm)
|
O.d. (mm)
|
Catalog Number
|
|
14/20
|
255
|
54
|
|
|
24/40
|
275
|
54
|
|
|
29/32
|
275
|
54
|
|
Back to top
Water cooled distilling heads
 |
Useful for removal of solvents from reaction mixtures or concentrations from solutions prior to crystallization.
|
S.T. Joints
|
Overall H (mm)
|
O.d. (mm)
|
Side-arm clearance (mm)
|
Catalog Number
|
|
14/20
|
210
|
35
|
110
|
|
|
24/40
|
400
|
54
|
225
|
|
|
29/32
|
400
|
54
|
225
|
|
|
Back to top
Multi-purpose distillation heads
 |
With vacuum take-off and anti-splash design. 4mm PTFE stopcock. (2mm PTFE stopcock for S.T. 14/20 version)
|
S.T. Joints
|
Overall H (mm)
|
O.d (mm)
|
Catalog Number
|
|
14/20
|
230
|
28
|
|
|
24/40
|
430
|
48
|
|
|
29/32
|
415
|
48
|
|
|
Back to top
Anti-creep distilling heads
 |
|
S.T. Joints
|
Overall H (mm)
|
Thermometer S.T Joint
|
Immersion (mm)
|
Catalog Number
|
|
14/20
|
140
|
10/18
|
40
|
|
|
19/22
|
140
|
10/18
|
40
|
|
|
24/40
|
225
|
10/30
|
75
|
|
|
29/32
|
225
|
10/30
|
75
|
|
|
Back to top
Distilling adapters
 |
|
S.T. Joints
|
Thermometer S.T. Joint
|
Immersion (mm)
|
Side-arm S.T. Joint
|
Catalog Number
|
|
14/20
|
10/18
|
40
|
150
|
|
|
24/40
|
10/30
|
75
|
150
|
|
|
24/40
|
10/30
|
75
|
225
|
|
|
29/32
|
10/30
|
75
|
225
|
|
|
Back to top
Short-path distillation apparatus
 |
With jacketed condenser.
|
S.T. Joints
|
Thermometer S.T. joint
|
Immersion (mm)
|
Condenser L (mm)
|
Catalog Number
|
|
14/20
|
10/18
|
40
|
150
|
|
|
24/40
|
10/30
|
75
|
150
|
|
|
24/40
|
10/30
|
75
|
300
|
|
|
29/32
|
10/30
|
75
|
300
|
|
|
Back to top
Short-path distillation head
 |
|
S.T. Joints
|
Overall H (mm)
|
Thermometer S.T. Joint
|
Immersion (mm)
|
Catalog Number
|
|
14/20
|
135
|
10/18
|
40
|
|
|
19/22
|
135
|
10/18
|
40
|
|
|
24/40
|
200
|
10/30
|
75
|
|
|
29/32
|
200
|
10/30
|
75
|
|
|
Back to top
Aldrich Claisen distillation adapters
 |
Claisen adapter with a thermometer joint. S.T. side-arm inner drip-tip joint.
|
S.T. Joints
|
Thermometer S.T. Joint
|
Immersion (mm)
|
Catalog Number
|
|
14/20
|
10/18
|
40
|
|
|
19/22
|
10/18
|
40
|
|
|
24/40
|
10/30
|
75
|
|
|
29/32
|
10/30
|
75
|
|
|
Back to top
Claisen distillation heads
 |
With Vigreux indents.
|
S.T. Joints
|
Overall H (mm)
|
Thermometer S.T. Joint
|
Immersion (mm)
|
Catalog Number
|
|
14/20
|
250
|
10/18
|
40
|
|
|
24/40
|
380
|
10/30
|
75
|
|
|
29/32
|
380
|
10/30
|
75
|
|
|
Back to top
Fractional distillation head
With Vigreux indents. With S.T. 14/20 and 24/40 male joints and S.T. 14/20 female joint. Overall H 300mm.
Z16,401-1
Back to top
Aldrich heavy-duty Vigreux, jacketed distillation head
 |
These distillation heads are designed with heavier tubing and extra support to withstand the rigors of laboratory use. The one-piece jacketed design achieves greater distillation efficiency.
|
S.T. Joints
|
Thermometer S.T. Joint
|
Immersion (mm)
|
O.D. (mm)
|
Condenser L (mm)
|
Catalog Number
|
|
14/20
|
10/18
|
40
|
25
|
150
|
|
|
24/40
|
10/30
|
75
|
38
|
300
|
|
|
29/32
|
10/30
|
75
|
38
|
300
|
|
|
Back to top
Micro distillation head apparatus
 |
One-piece assembly Vacuum jacketed head with attached condenser. Has Vigreux indents and thermowell for 40mm immersion thermometer. S.T. 14/20M joints and S.T. 10/18F joints.
Z22,010-8
Two-piece assembly As above, but with S.T. 14/20M joint connection between head and condenser.
|
Back to top
Fraction cutter
 |
50mL barrel, 4mm glass stopcocks
|
S.T. Joints
|
Overall H (mm)
|
Catalog Number
|
|
24/40
|
300
|
|
|
29/32
|
300
|
|
|
Back to top
Micro distillation head
 |
 |
S.T. 24/40 joint and 2mm glass stopcock with O-ring. Use with recovery flask below.
Z12,960-7
Recovery flask, 100mL
Z51,557-4 |
Back to top
Quick-distillation and sample collection tube
S.T. 35/45 joint, S.T. 10/18 thermometer joint. For distilling small samples.
Z14,796-6
Back to top
Variable take-off splitter heads
 |
|
|
Micro distilling heads
The column and receiver joints are S.T. 14/20 with a S.T. 10/18 thermometer joint, two 2mm glass stpococks, PTFE 0-3 mm metering valve.
|
Description
|
Overall H (mm)
|
Immersion (mm)
|
Catalog Number
|
|
Jacketed
|
260
|
40
|
|
|
Unjacketed
|
260
|
40
|
Standard distilling heads
The column and receiver joints are S.T. 24/40 with a S.T. 10/30 thermometer joint, two 4mm glass stopcocks, PTFE 0-3 mm metering valve.
|
Description
|
Overall H (mm)
|
Immersion (mm)
|
Catalog Number
|
|
Jacketed
|
490
|
75
|
|
|
Unjacketed
|
490
|
75
|
|
|
Jacketed
|
490
|
40
|
|
| S.T. 29/32 with a S.T. 10/19 thermometer joint. |
|
Back to top
High-capacity variable take-off splitter head
 |
One-piece jacketed design features a heavy-duty condenser and jacketed drip-tip joint. The column and receibers joints ST. 24/40 with a S.T. 10/30 thermometer joint. Two 4mm glass stopcocks, PTFE 0-3 mm metering valve
|
|
|
Overall H (mm)
|
Immersion (mm)
|
Catalog Number
|
|
510
|
75
|
|
|
Back to top
Aldrich falling-film distillation heads
|
The material is in contact with the heated center finger for only a brief period of time, avoiding decomposition by long-term exposure to heat. This system can be used at atmospheric or reduced pressure. Fig. 1 shows a typical distillation set-up.
|
Falling Fi | | | |