PACKING
On this page:
Aldrich glass helices
PRO-PAK distillation packings
Solid-borosilicate glass beads
PTFE boiling stones
Aldrich glass helices
 |
Each helice consists of a single
turn of borosilicate glass rod, approximately 0.5mm in diam.
|
Approx. coil i.d. (mm)
|
Catalog Number
|
|
2
|
|
|
3
|
|
|
4-5
|
|
|
6
|
|
|
Back to top
PRO-PAK distillation packings
Efficient,
self-wetting, protruded metal dump-packing for distillation columns of
0.75 to 12in. diameter. Maximum reflux rate of approximately 3.4L/h. Free
space for 0.16in.2 size is 94% and for 0.24in.2
size is 96%. Each package has a packing volume of approx. 1L.
|
Metal
|
0.16in.2
Catalog Number
|
0.24in.2
Catalog Number
|
| 316 SS |
|
|
| Monel |
|
|
Back to top
Solid-borosilicate glass
beads
 |
For use as distillation column packing, mixing
beads, and boiling stones. Each 454g box has a packing volume of
approx. 360cm3.
|
Diam.
(mm)
|
Catalog
Number
|
|
1
|
|
|
2
|
|
|
3
|
|
|
4
|
|
|
5
|
|
|
6
|
|
|
Back to top
PTFE boiling stones
 PTFE
stones are not affected by boiling acids or alkalis, ketones, esters,
or alcohols. Easily cleaned and will not scratch glassware.
Z24,355-8
Back to top |