|
// We only want a grey bar if c3_headline has a value...
$headline="";
if ($headline) {
print " | ";
} else {
print " | ";
}
?>
|
Apparatus
Columns
Packing
Heads
Traps
|
TRAPS
Dean-Stark Traps Cold-Traps
& Vacuum Traps
Dean-Stark Traps
On this page:
Dean-Stark traps
Jacketed Dean-Stark trap
Modified Dean-Stark apparatus
Modified, jacketed Dean-Stark trap
"N"-tubes
Dean-Stark traps
 |
Designed specifically for azeotropic
distillation using solvents lighter than water. Each Dean-Stark trap
comes standard with a 4mm bore PTFE stopcock. The 12mL and 25mL traps
use a 2mm bore stopcocks.
 |
|
Cap.
(mL)
|
Height
(mm)
|
Side-arm
clearance (mm)
|
Suggested
flask size
|
S.T.
14/20 Joints
Catalog Number
|
S.T.
24/40 Joints
Catalog Number
|
S.T.
29/32 Joints
Catalog Number
|
|
12
|
205
|
80
|
50mL - 5L
|
|
-
|
-
|
|
25
|
255
|
80
|
50mL - 5L
|
|
-
|
-
|
|
25
|
280
|
80
|
50mL - 5L
|
-
|
100
|
350
|
100
|
5L - 12L
|
-
|
300
|
440
|
170
|
22L - 50L
|
-
|
500
|
455
|
170
|
22L - 50L
|
-
|
|
|
Back to top
Jacketed Dean-Stark trap
For azeotroping large amounts of water, 325mL capacity. With S.T. 24/40 joints
and 4mm bore PTFE stopcock.
Z10,257-1
Back to top
Modified Dean-Stark apparatus
|
|
The 8mL reservoir version is specifically designed
for small scale azeotropic distillation of valuable solvents using
heavy water. 2mm bore PTFE stopcock, 300mm overall height.
|
 |
The modified Dean-Stark technique
(See Fig. 1)
- Solvent vapors condense and collect in the receiver
- The modified Dean-Stark trap separates the two components
- The heavier than water layer sinks to the bottom of the trap
leaving the water on top
- The solvent is forced back through the inside tube and returned
to the boiling flask
- The upper water layer is drained out through the 2mm PTFE stopcock
|
|
Reservoir
cap. (mL)
|
S.T.
Joint 24/40
Catalog Number
|
S.T.
Joint 29/32
Catalog Number
|
|
8
|
35
|
|
|
Back to top
Modified, jacketed Dean-Stark
trap
 |
|
350mL capacity. 0-3mm PTFE valve.
300mm overall height.
Z24,960-2 |
Back to top
"N"-tubes
 |
For small-scale distillation.
10mL capacity.
|
Description
|
Catalog
Number
|
| S.T. 19/38F, 24/40M
joints |
|
| S.T.
24/40 joints |