|
// We only want a grey bar if c3_headline has a value...
$headline="";
if ($headline) {
print " | ";
} else {
print " | ";
}
?>
|
Miscellaneous
Thimbles
Systems
|
MISCELLANEOUS EXTRACTORS
On this page:
Aldrich convertible liquid/liquid continuous extractors
Aldrich continuous extractors Ace soxhlet extraction
apparatus
Aldrich convertible liquid/liquid continuous extractors
|
|
For solvents
heavier than water, stopcock is open and A is not used. For
solvents lighter than water, stopcock is closed and A is
inserted into B; condensate drips into and through diffuser
tube A to the bottom of the extractor. PTFE-stopcock.
|
|
Cap. (mL)
|
S.T.
Joint (B)
|
S.T.
Joint (C)
|
Overall
H
(mm)
|
Complete
Catalog Number
|
Replacement
part (A)
Catalog Number
|
|
100
|
24/40
|
24/40
|
400
|
250
|
24/40
|
24/40
|
420
|
500
|
24/40
|
24/40
|
430
|
1,000
|
55/50
|
24/40
|
635
|
2,000
|
55/50
|
24/40
|
735
|
4,000
|
55/50
|
29/32
|
815
|
|
|
Back to top
Aldrich continuous extractors
 |
S.T. top and bottom joints with a flat flange on
the removable top. Separate inner glass tube has a 37mm perforated
bottom for support of filter paper.
|
|
S.T.
Joint
|
Overall
H (mm)
|
Catalog
Number
|
|
24/40
|
430
|
|
|
29/32
|
430
|
|
| Replacement
clamp |
|
|
Back to top
Ace soxhlet extraction
apparatus
The improved design of Ace extractor condensers permits
greater condensing capabilities. Cycling rates may be doubled over conventional
extractors. Apparatus includes extractor body, condenser, and flask.
|
Flask
cap.
(mL)
|
Extractor
cap. (mL)
|
Thimble
size (mm)
|
Catalog
Number
|
|
125
|
50
|
25 x 80
|
|
|
250
|
85
|
33 x 80
|
|
|
300
|
145
|
33 x 94
|
|
|
300
|
200
|
43 x 123