|
Glassware Catalog
Flask Accessories
|
Filtration • Flat-Bottom • Round-Bottom • Multi-Neck • Schlenk • Septum-Inlet • Accessories
|
On this page: The Aldrich flask throne • BIBASE rubber support rings • Cork flask support rings
The Aldrich flask throne
|
|
Provides safe, hands-free support for round-bottom flasks sizes 10 to 22L. A slotted hole provides secure support while tipping flask joint downward to dump contents. 19 L x 19 W x 10in. H.
- Prevents flask breakage, loss of product, and chemical exposure
- For benchtop use or inside fume hoods to transfer product from flask to bottles or trays
- Pays for itself with the cost of just one broken flask
- Chemical resistantLDPE
Z41,156-6
|
Back to top
BIBASE rubber support rings
 |
Reversible; can also be stacked upon each other. Three sizes provide six different diameters for round-bottom flasks ranging from 50 to 6,000mL. Excellent adhesion to glass prevents slippage and also offers thermal and electrical insulation. Available in two rubber types.
Black natural rubber • usable at temperatures from -20 to 70°C (dry heat), -20 to 120°C (moist heat) and has excellent chemical resistance.
Blue silicone • usable at temperatures from -20 to 200°C. Withstands repeated high temperature or radiation. Non-toxic, biologically neutral.
|
|
Flask size (mL)
|
Black natural rubber Catalog Number
|
Blue silicone Catalog Number
|
|
50 - 500
|
500 - 2,000
|
2,000 - 6,000
|
|
|
Back to top
Cork flask support rings
 |
With internal bevel.
|
Size (mm) I.d. x o.d.
|
Catalog Number
|
|
30 x 80
|
|
|
60 x 110
|
|
|
90 x 140
|
|
|
120 x 170
|
|
|
150 x 210
|
|
|
180 x 230