Sigma-Aldrich
Labware logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Glassware Catalog > Schlenk Flasks
Glassware Catalog
Schlenk Flasks
Filtration  •  Flat-Bottom  •  Round-Bottom  •  Multi-Neck  •  Schlenk  •  Septum-Inlet  •  Accessories
On this page:
Kjeldahl reaction and storage flasksReaction and storage tubesSolvent storage flasks
Schlenk storage vessels have a 2mm glass stopcock. The side-arm accommodates a septum (use Z10,072-2 or Z12,435-4).
al_glassware_note84
Kjeldahl reaction and storage flasks
al_glassware_flasks3
Cap. (mL)
S.T. Joint
Outer joint
Catalog Number
Inner joint
Catalog Number
25
14/20
50
14/20
100
14/20
100
24/40
100
29/32
250
14/20
250
24/40
250
29/32
See the Vacuum Equipment Section for
Vacuum manifolds with S.T. Joints
Back to top
Reaction and storage tubes
al_glassware_flask4
Cap. (mL)
S.T. Joint
Outer joint
Catalog Number
Inner Joint
Catalog Number
25
14/20
50
14/20
100
14/20
100
24/40
100
29/32
250
14/20
250
24/40
250
29/32
Back to top
Solvent storage flasks
al_Z51586L2
Cap. (mL)
S.T. joint
Catalog Number
25
14/20
50
14/20
100
14/20
100
24/40
100
29/32
250
14/20
250
24/40
250
29/32
500
24/40
500
29/32
1,000
24/40
1,000
29/32
Back to top