All
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Substructure
YFG Search
Labware Products
Learning Center
Custom Glassware
Corning Products
Gadgets
Glassware Catalog
Ordering
Labware Literature
SciBookSelect
Email this page to a friend
To Email:
From Email:
Message:
Labware
>
Glassware Catalog
>
Buchner Funnels
Glassware Catalog
Buchner Funnels
Addition
• Buchner •
Filter/Drying
•
Miscellaneous
On this page:
Aldrich constant-temperature Buchner funnel
Aldrich vacuum-jacketed Buchner funnel
•
Ace Buchner funnels
Aldrich constant-temperature Buchner funnel
With jacket for circulating steam or coolant. 60mL capacity, 180mm H, 40mm diam. frit, 25 to 50 µm porosity. Drip-tip joint and vacuum take-off.
S.T. joint
Catalog Number
24/40
Z17,583-8
29/32
Z51,775-5
Back to top
Aldrich vacuum-jacketed Buchner funnel
Silvered, 60mL capacity, 180 mm H, with 40mm diam. frit, 25 to 50 µm porosity. Drip-tip joint and vacuum take-off.
S.T. joint
Catalog Number
24/40
Z17,581-1
29/32
Z51,776-3
Back to top
Ace Buchner funnels
With glass frit, hose connection, and S.T. 24/40 joint.
Porosity (µm)
Catalog Number
Disc diam.: 40mm; capacity: 60mL
145 - 175
Z12,579-2
70 - 100
Z12,580-6
25 - 50
Z12,581-4
10 - 20
Z12,582-2
4 - 8
Z12,583-0
Disc diam.: 65mm; capacity: 140mL
145 - 175
Z15,808-9
70 - 100
Z15,810-0
25 - 50
Z15,812-7
10 - 20
Z15,814-3
4 - 8
Z15,817-8
Disc diam.: 80mm; capacity: 350mL
145 - 175
Z15,809-7
70 - 100
Z15,811-9
25 - 50
Z15,813-5
10 - 20
Z15,815-1
4 - 8
Z15,818-6
Back to top
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.