Sigma-Aldrich
Labware logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Glassware Catalog > Miscellaneous Funnels
Glassware Catalog
Miscellaneous Funnels
Addition  •  Buchner  •  Filter/Drying  •  Miscellaneous
On this page:
Aldrich gas reducing flasksAldrich rapid transfer funnelsLarge separatory funnels
Aldrich gas reducing flasks
al_Z21444L
Features a large frit, porosity C, for efficient gas dispersion. Flasks have a
S.T. 14/20 threaded side inlet joint.
Cap.(mL) S.T. Threaded
Top joint
Frit diam. (mm) Diam x H (mm) Catalog Number
250 24/40 40 45 x 470 Z21,444-2
1,000 45/50 65 75x 540 Z21,445-0
Back to top
Aldrich rapid transfer funnels al_glassware_note87
al_Z54884L
140mm diameter, 250mm height, 3.5mm wall thickness.
Ball joint Catalog Number
65/40 Z54,884-7
75/50 Z54,885-5
Back to top
Large separatory funnels
al_Z12784L
10mm bore glass stopcock and 50mm (85mm on 22L) top opening. Allows agitation with "Lightnin"-type stirrer.
Cap. (L) Catalog Number
12 Z10,260-1
22 Z10,262-8
PTFE stopcock and S.T.45/40 joint.
Cap. (L) Catalog Number
4 Z12,784-1
6 Z12,786-8
10 Z12,787-6
S.T. 45/40 Glass pennyhead stopper Z22,970-9
Back to top