|
// We only want a grey bar if c3_headline has a value...
$headline="";
if ($headline) {
print " | ";
} else {
print " | ";
}
?>
|
Glas-Col Series O
Glas-Col Series TM
Clas-Col Controllers
|
|
GLAS-COL SERIES O HEATING MANTLES
On this page:
Hemispherical heating mantles Buchner
funnel mantles
Series O mantles are the original and the most popular heating mantles.
- Made from flexible fabrics
- Fit easily into tight spaces
- Maximum 450°C internal operating temperature
- CSA and CE mark of conformity
All mantles include a 4ft, 2-wire cord and locking connector. A temperature
controller is required for operation.
Hemispherical heating mantles
The mantles cover the bottom half of single and multi-neck flasks, allowing
for full view of contents.
|
Flask
size (mL)
|
Watts
|
115V
Catalog Number
|
230V
Catalog Number
|
|
50
|
60
|
|
-
|
|
100
|
80
|
|
-
|
|
125
|
80
|
|
-
|
|
200
|
100
|
|
-
|
|
250
|
180
|
300
|
180
|
|
-
|
|
500
|
270
|
1,000
|
380
|
2,000
|
500
|
3,000
|
500
|
|
-
|
|
5,000
|
600
|
12,000
|
2 @ 650
|
|
-
|
Back to top
Buchner funnel mantles
 |
- Provides gentle heat for solutions that must be kept warm during transfer
- Not CE compliant
|
|
Top diam.
(in.)
|
Coors
Funnel No.
|
Watts
|
115V
Catalog Number
|
230V
Catalog Number
|
|
12.56
|
60240
|
50
|
|
-
|
|
13.38
|
60242
|
50
|
|
-
|
|
14.06
|
60243
|
70
|
|
-
|
|
14.81
|
60244
|
70
|
|
-
|
|
15.38
|
60245
|
100
|
16.50
|
60246
|
220
|
18.13
|
60247
|
300
|
10.5
|
60248
|
350