|
// We only want a grey bar if c3_headline has a value...
$headline="";
if ($headline) {
print " | ";
} else {
print " | ";
}
?>
|
LATTICE SYSTEMS
On this page:
Aldrich Lattice System
Sets Aldrich lattice rods Mounting
adapters Channel clamp
Lattice-rod clamps
"T"-handle screws
Quick-action lattice-rod clamps
Base plates
Sliding-extension-clamp couplings
Extension coupling
Aldrich Custom Lattice
Systems
|
|
- Pre-cut lattice-rods and hardware are available from stock
for immediate shipment
- Custom-length lattice-rods can be ordered from Aldrich
- Special pricing for bulk orders of rods and hardware
Please contact your local Sigma-Aldrich Location
for a quotation on custom-length lattice rods or bulk orders.
Back to top
|
Lattice System Hardware
Aldrich lattice rods
Rods are 1/2in. diameter.
|
L (in.)
|
Fiberglass
Catalog Number
|
Aluminum
Catalog Number
|
316
SS
Catalog Number
|
|
8
|
|
-
|
-
|
|
12
|
|
-
|
-
|
|
24
|
30
|
|
-
|
-
|
|
34
|
|
-
|
-
|
|
44
|
|
-
|
-
|
|
48
|
72
|
96
|
|
-
|
|
|
108
|
-
|
-
|
|
|
144
|
-
|
-
|
|
Back to top
All hardware is for 1/2in. diameter
lattice rods.
Mounting
adapters
|
For mounting lattice-support rods to benchtops already equipped
with 1/2in. countersunk fittings. Aluminum with SS set screws 1/2in.
NPTM thread.
Z12,498-2
|
Back
to top
|
|
Channel clamp
|
For fastening lattice-support
rods to the channel frames in Aldrich lattice systems
above. Can be loosened from frame to position rods.
Z12,516-4
|
Back to top
|
|
Lattice-rod clamps
Back to top
|
Die-cast aluminum alloy clamp equipped
with Allen set screws. Assembles without dismantling the existing
lattice-rod setup.
Z10,717-4 |
"T"-handle screws
Back to top
|
Converts lattice-rod clamp (Z10,717-4)
to quick-action clamp (Z10,858-8).
Z10,859-6 |
Quick-action lattice-rod clamps
|
Back to top
|
Similar to above, but equipped
with "T"-handle and SS screws, permitting rod adjustment
without tools.
Z10,858-8 |
Base plates
Back to top
|
Mounts lattice-support rods to benchtop, wall,
or ceiling. Unthreaded type provides perpendicular support. Swivel
type permits locking a rod in any position up to 22.5° from
vertical using two Allen screws set at right angles to each other.
|
Description
|
Catalog
Number
|
|
Unthreaded type
|
|
|
Swivel type
|
|
|
| Sliding-extension-clamp
couplings |
Back to top
|
38mm L, with thumbscrews. Connects
lattice rods
side-by-side for length adjustments.
Z19,325-9
|
Extension coupling
Back to top
|
76mm L, with thumbscrews. Connects
lattice rods end-to-end.
Z19,324-0 |