|
// We only want a grey bar if c3_headline has a value...
$headline="";
if ($headline) {
print " | ";
} else {
print " | ";
}
?>
|
NMR Tubes
Aldrich NMR Tube Cleaners
Specialty NMR Tube Cleaners
NMR Tube Manifolds
|
NMR TUBE MANIFOLDS AND ACCESSORIES
On this page:
Aldrich 5mm NMR tube filling manifolds Aldrich
5mm NMR tube evacuation manifolds
5mm NMR tube with PTFE valve NMR-tube
Dewar flask Pocket NMR tube carriers
Aldrich 5mm NMR tube filling manifolds
NMR Tube Filling Technique
- Insert NMR tube through
the open top cap and Viton O-ring (see figure 1)
- Evacuate NMR tube
- Fill NMR tube with a syringe
through the PTFE-faced silicone septa (see figure 2)
- Tip-off NMR tube or remove
tube and cap (see figure 3)
|
 |
Manifolds have 0-10mm bore PTFE
valves. The multiple filling station units allow removal of one
NMR tube while the remaining tube(s) stay under vacuum.
|
Filling
Stations
|
L
(mm)
|
Catalog
Number
|
|
1
|
100
|
|
|
2
|
150
|
|
|
5
|
300
|
|
|
Replacement
Parts
|
Description
|
Catalog
Number
|
| Replacement
PTFE-faced silicone septa |
|
| Replacement
screw caps |
|
| Replacement
Viton O-rings |
|
|
Accessories
|
|
|
| For
vacuum-thaw applications, use the Aldrich NMR tube Dewar flask.
|
When
the tip-off technique is used, the MT-51
butane torch (listed below) generates a 1,300°C pinpoint flame.
|
|
Description
|
Catalog
Number
|
| NMR tube Dewar flask |
|
| MT-51
handheld torch |
|
|
(Torch shipped empty, order butane below) |
| Butane
torch fuel, 26g of butane fuel per can |
|
Back to top
Aldrich 5mm NMR tube evacuation
manifolds
Manifolds have 0-10mm bore PTFE valves. The multiple evacuation station
units allow removal of one NMR tube while the remaining tube(s) stay under
vacuum.
|
Evacuation
stations
|
L (mm)
|
Catalog
Number
|
|
1
|
100
|
|
|
2
|
150
|
|
|
5
|
300
|
|
Back to top
Accessories
5mm NMR tube with PTFE valve
Improve the handiling of NMR samples. A convenient alternative to flame-sealing.
Z18,397-0
Back to top
NMR-tube Dewar flask
Compact
size minimizes volume of liquid nitrogen required. Borosilicate glass
with protective aluminum housing. 2.0 i.d. x 23cm depth.
Z24,729-4
Back to top
Pocket NMR tube carriers
 |
Unbreakable NMR tube carrier fits into lab-coat
pocket like a pen. Has a snap-on cap and a shirt clip that secures
carrier in pocket. Holds one 5mm diam. x 7in. L NMR tube.
|
Color
|
Catalog
Number
|
| Red |
|
| White |
|
| Blue |
|
| |
| Color
assortment pack |
|
| One
of each color carrier; red, white and blue |