|
| |
|
NMR Tubes
Aldrich NMR Tube Cleaners
Specialty NMR Tube Cleaners
NMR Tube Manifolds
|
NMR TUBES
On this page:
Selection Guide
NMR Tubes
Shigemi NMR Microtube Assemblies
- Made of ASTM Type I borosilicate glass* for compatibility with ground joints, valves,
and vacuum racks
- Precision ground and highly polished for spinning stability and highest resolution
- Thin-wall glass with round- bottoms
- Sandblasted marking spot for labeling samples
*Except disposable grade NMR tubes.
NMR Tube Selection Guide
Choose the NMR tube grade that most closely matches the frequency (in MHz)
of the spectrometer.
| Aldrich
Grade |
MHz
|
Applications
|
Cross
Reference No.
(5mm tubes) |
| Wilmad
|
Kontes
|
|
Disposable
|
30
|
Student
labs, basic studies
|
WG-Economy
|
-
|
|
Series
30
|
30
|
Student
labs, basic studies
|
505-PS
|
897200
|
|
Series
200
|
60
to 270
|
Lower
field studies
|
506-PP |
897205 |
|
Series
300
|
60
to 360
|
Routine
applications
|
507-PP
|
897220
|
|
Series
400
|
200
to 500
|
High-field,
multiple-scan FT-NMR
|
528-PP |
897235 |
|
Series
500
|
500+
|
Ultra-high
resolution
|
535-PP
|
897240
|
|
Series 800
|
600
to 800
|
Highest
resolution and performance
|
541-PP |
897245 |
Back to top
NMR Tubes
| Grade
|
L
(in.) |
Wall
uniformity (TIR) |
Camber
(in.) |
Catalog
Number |
| 3mm Microsample NMR tubes
|
|
Series 300
|
7
|
0.0020
|
0.0010 |
Z41,290-2
|
| |
8
|
0.0020
|
0.0011 |
Z41,291-0
|
|
Series 400
|
7
|
0.0010
|
0.0005 |
Z41,292-9
|
| |
8 |
0.0010 |
0.0006 |
Z41,293-7
|
|
Series 500
|
7
|
0.0005
|
0.00025
|
Z41,294-5
|
| |
8 |
0.0005 |
0.00035
|
Z41,295-3
|
|
5mm NMR tubes
|
|
Disposable
|
7 |
-
|
-
|
Z41,277-5
|
|
Series 30
|
7 |
0.0030 |
0.002 |
Z41,278-3
|
|
Series 200
|
7
|
0.0025
|
0.002 |
Z41,279-1
|
|
Series 300
|
7 |
0.0020 |
0.0010 |
Z41,280-5
|
| |
8
|
0.0020
|
0.0011 |
Z41,281-3
|
| |
9 |
0.0020 |
0.0012 |
Z41,282-1
|
|
Series 400
|
7
|
0.0010
|
0.0005 |
Z41,284-8
|
| |
8 |
0.0010 |
0.0006 |
Z41,285-6
|
|
Series 500
|
7
|
0.0005
|
0.00025
|
Z41,286-4
|
| |
8 |
0.0005 |
0.00035
|
Z41,287-2
|
|
Series 800
|
7
|
0.0002
|
0.00015
|
Z41,288-0
|
| |
8 |
0.0002 |
0.00015
|
Z41,289-9
|
| 10mm NMR tubes
|
|
Series 200
|
7
|
0.0030
|
0.0015 |
Z41,296-1
|
|
Series 400
|
7 |
0.0015 |
0.0005 |
Z41,298-8
|
TIR-Total Indicator Reading
|
For
more information, please contact your local sales office.
|
Back to top
Shigemi NMR Microtube Assemblies
These microtube assemblies consist of a small volume inner microtube mounted
in a standard size NMR tube. When making measurements with these tubes,
the small volume of sample solution is held in the region of the NMR tube
length that the NMR coil "sees", without large portions of the
sample solution being excluded from the measurement. In addition, the
assemblies are magnetic susceptibility matched to four common NMR solvents;
D2O, CD3OD, CDCl3 and DMSO.
Small volumes mean it is possible to use increased concentration sample
solutions, leading to measurements with improved signal-to-noise ratios
and greatly reduced spectrum acquisition times. The solvent matching of
these tubes means the small sample size has no significant effect on spectral
resolution or sideband.
Protein studies use small sample sizes and low concentration solutions.
Shigemi microtubes are ideal for small samples, allowing improved concentrations
and they offer great cost savings through reduced spectrometer run-time
and quickly pay back the additional expense in buying these specialist
microtubes.
-
Magnetic susceptibility matched to a variety of solvents for the
best resolution in these solvents using a minimal amount of sample
-
The small sample size will not affect the spectral resolution or
the sidebands
-
Each microtube assembly consists of an insert and an outer sample tube
Back to top
5mm Symmetrical NMR microtubes
Tube insert : 190.0 L x 4.1mm o.d., outer sample tube: 180.0 x 4.965mm.
*Choose the bottom insert length that matches the probe coil most closely.
|
Shigemi No. |
Bottom insert L* (mm) |
Catalog Number |
Tubes matched with CDCl3 |
|
CMS-005B
|
8 |
Z52,934-6 |
|
CMS-005J
|
12 |
Z52,935-4 |
|
CMS-005V
|
15 |
Z52,936-2 |
|
CMS-CAP
|
Symmetrical microtube
cap |
Z52,937-0 |
|
Tubes matched with DMSO
|
|
DMS-005B
|
8 |
Z52,938-9 |
|
DMS-005J
|
12 |
Z52,939-7 |
|
DMS-005V
|
15 |
Z52,940-0 |
|
DMS-CAP
|
Symmetrical microtube
cap |
Z52,941-9 |
|
Tubes matched with D2O
|
|
BMS-005B
|
8 |
Z54,334-9 |
|
BMS-005J
|
12 |
Z52,942-7 |
|
BMS-005V
|
15 |
Z52,943-5 |
|
BMS-CAP
|
Symmetrical microtube
cap |
Z52,944-3 |
Back to top
|
|
|
Advanced NMR microtubes
-
Thinner glass wall in the sample area than symmetrical microtube
(0.20mm vs 0.40mm), increasing the sample capacity held in the Rf
region by a factor of 1.2
-
16% better signal to noise ratio than symmetric tubes
-
Despite the increased sample volume, artifacts caused by the Rf inhomogeneity
is very limited and increased sensitivity is expected
Specifications:
- 5mm o.d. tubes: insert : 190.0 L x 4.1mm o.d.; outer sample tube:
180.0 x 4.965mm.
- 8mm o.d. tubes: insert : 200.0 L x 6.9mm o.d.; outer sample tube:
190.0 x 8.0mm.
*Choose the bottom insert length that matches the
probe coil most closely.
|
Shigemi No. |
Outer tube o.d. (mm) |
Bottom insert L* (mm) |
Catalog Number |
Tubes matched with CDCl3 |
|
CMS-005TB
|
5 |
8 |
Z52,908-7 |
|
CMS-005TJ
|
5 |
12 |
Z52,909-5 |
|
CMS-005TV
|
5 |
15 |
Z52,910-9 |
|
CMS-008TB
|
8 |
8 |
Z52,911-7 |
|
CMS-008TJ
|
8 |
12 |
Z52,912-5 |
|
CMS-008TV
|
8 |
15 |
Z52,913-3 |
|
Tubes matched with CD3OD
|
|
MMS-005TB
|
5 |
8 |
Z52,914-1 |
|
MMS-005TJ
|
5 |
12 |
Z52,916-8 |
|
MMS-005TV
|
5 |
15 |
Z52,917-6 |
|
MMS-008TB
|
8 |
8 |
Z52,918-4 |
|
MMS-008TJ
|
8 |
12 |
Z52,919-2 |
|
MMS-008TV
|
8 |
15 |
Z52,920-6 |
|
Tubes matched with DMSO
|
|
DMS-005TB
|
5 |
8
|
Z52,921-4 |
|
DMS-005TJ
|
5 |
12 |
Z52,922-2 |
|
DMS-005TV
|
5 |
15
|
Z52,923-0 |
|
DMS-008TB
|
8 |
8 |
Z52,924-9 |
|
DMS-008TJ
|
8 |
12
|
Z52,925-7 |
|
DMS-008TV
|
8 |
15 |
Z52,926-5 |
|
Tubes matched with D2O
|
|
BMS-005TB
|
5 |
8 |
Z52,945-1 |
|
BMS-005TJ
|
5 |
12
|
Z52,927-3 |
|
BMS-005TV
|
5 |
15 |
Z52,928-1 |
|
BMS-008TB
|
8
|
8
|
Z52,930-3 |
|
BMS-008TJ
|
8 |
12 |
Z52,931-1 |
|
BMS-008TV
|
8
|
15
|
Z52,933-8 |