ALDRICH NMR TUBE CLEANERS
INTEGRAL SOLVENT
RESERVOIR (Figure 1)
Models A, B, C
A PP tube cap placed on the closed end of an inverted NMR tube holds
the tube in place and seals the system to allow efficient vacuum. Applied
suction sprays solvent inside and against the closed end of tube for
thorough cleaning.
Model D
As above, except the ground-glass stopper is the cap.
|
Model
|
S.T. Joint
|
For tube
(mm)
|
Catalog
Number
|
|
A
|
Tooled end
|
5
|
|
|
B
|
24/40
|
5
|
29/32
|
5
|
24/40
|
10
|
|
|
C
|
24/40
|
5
|
29/32
|
5
|
24/40
|
10
|
|
|
D
|
24/40
|
5
|
|
Reservoir flask (1,000mL) with hose barb |
| |
24/40
|
|
|
| |
29/32