Order Center
Equipment, Supplies & Books
Corning Products
Product Lines
Products by Application
Product Highlights
Books
Gases Equipment
Glassware Catalog
Key Resources
Labware Literature
New Products
"; } else { print ""; } ?>
SUBLIMATION
On this page:
Micro sublimation apparatusSublimation apparatusModified sublimation apparatus
Micro sublimation apparatus
S.T. 24/40 joint, 2mm glass stopcock with O-ring.
Z12,959-3
Recovery flasks for Z12,959-3
100mL
Z51,557-4
Back to top
Sublimation apparatus
Flat-bottom on inner tube for easy sample removal. 2mm glass stopcock.
Starting material capacity
Catalog Number
Up to 3g
Up to 12g
Back to top
Modified sublimation apparatus
Removable vacuum inlet with 2mm glass stopcock improves handling during sample removal and reduces breakage of glassware.
Starting material capacity
Catalog Number
Up to 20g